Documents
Z761001
TPP® easy read cell counter
Synonym(s):
Automated cell counter, Cell counting device
About This Item
UNSPSC Code:
42181504
NACRES:
NB.22
packaging
pack of 1 ea
manufacturer/tradename
TPP 87010
measuring range
0-5 μL
General description
Features of the "easy read" mesuring device:
  • Reading device with magnifying glass and cross line
  • Optimal usage in combination with the PCV tube
  • Reproducible cell pellet measuring possible
  • Reusable product, no further investments necessary
  • Measuring range : 0 - 5 μl
Legal Information
TPP is a registered trademark of TPP Techno Plastic Products AG