Skip to Content
Merck
All Photos(1)

Documents

SML0295

Sigma-Aldrich

Dihydromyricetin

≥98% (HPLC)

Synonym(s):

(2R,3R)-3,5,7-Trihydroxy-2-(3,4,5-trihydroxyphenyl)-2,3-dihydrochromen-4-one, 3,3′,4′,5,5′,7-Hexahydroxyflavanone, Ampelopsin, Ampeloptin, DHM

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12O8
CAS Number:
Molecular Weight:
320.25
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

white to beige

solubility

DMSO: ≥5 mg/mL (warmed)

storage temp.

−20°C

SMILES string

O=C1C2=C(O)C=C(O)C=C2O[C@H](C3=CC(O)=C(C(O)=C3)O)[C@H]1O

InChI

1S/C15H12O8/c16-6-3-7(17)11-10(4-6)23-15(14(22)13(11)21)5-1-8(18)12(20)9(19)2-5/h1-4,14-20,22H/t14-,15+/m0/s1

InChI key

KJXSIXMJHKAJOD-LSDHHAIUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dihydromyricetin is a major flavonoid present in A. grossedentata.

Application

Dihydromyricetin has been used to study its effect on adipogenesis and glucose uptake in differentiated 3T3-L1 pre-adipocytes. It has also been used to study its antitumor activity against liver cancer cells.

Biochem/physiol Actions

Dihydromyricetin (Ampelopsin) is a flavanonol with antioxidant and anti-cancer activity, found to have anti-alcohol intoxication effects. Its anti-alcohol effects appear to be by its actions as a positive modulator of GABA-A receptors at the benzodiazepine site.
Dihydromyricetin possesses anti-inflammatory action.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Scott A Snyder et al.
Nature, 474(7352), 461-466 (2011-06-24)
Although much attention has been devoted to resveratrol, a unique polyphenol produced by plants and credited as potentially being responsible for the 'French paradox'--the observation that French people have a relatively low incidence of coronary heart disease, even though their
Jun Zhou et al.
Journal of separation science, 34(2), 160-168 (2011-01-20)
A simple, rapid and specific method based on cloud-point extraction (CPE) was developed to determine ampelopsin in rat plasma after oral administration by reversed-phase high-performance liquid chromatography. The non-ionic surfactant Genapol X-080 was chosen as the extract solvent. Some important
Li-Ping Ruan et al.
Journal of pharmaceutical and biomedical analysis, 38(3), 457-464 (2005-06-01)
The aim of this study was to increase the solubility of ampelopsin (AMP) in water by two systems: solid dispersions with polyethylene glycol 6000 (PEG 6000) or polyvinylpyrrolidone K-30 (PVP K30) and inclusion complexes with beta-cyclodextrin (BCD) and hydroxypropyl-beta-cyclodextrin (HPBCD).
Baolai Zhang et al.
Anti-cancer drugs, 23(6), 590-596 (2012-01-14)
The aim of this study was to establish xenograft models of tumor in mice bladder and evaluate the antitumor efficacy of ampelopsin sodium (Amp-Na). A total of 2×10 human bladder carcinoma EJ cells and murine sarcoma 180 cells were instilled
Shimei Qi et al.
International immunopharmacology, 12(1), 278-287 (2011-12-24)
Ampelopsin (AMP), a plant flavonoid, has potent anti-inflammatory properties in vitro and in vivo. The molecular mechanisms of ampelopsin on pharmacological and biochemical actions of RAW264.7 macrophages in inflammation have not been clearly elucidated yet. In the present study, non-cytotoxic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service