Skip to Content
Merck
Dihydromyricetin ≥98% (HPLC)

Dihydromyricetin

Dihydromyricetin ≥98% (HPLC)
Synonym(s):
(2R,3R)-3,5,7-Trihydroxy-2-(3,4,5-trihydroxyphenyl)-2,3-dihydrochromen-4-one, 3,3′,4′,5,5′,7-Hexahydroxyflavanone, Ampelopsin, Ampeloptin, DHM, (+)-Ampelopsin, (+)-Dihydromyricetin, (2R,3R)-3,5,7-trihydroxy-2-(3,4,5-trihydroxyphenyl)-2,3-dihydrochromen-4-one
Empirical Formula (Hill Notation):
C15H12O8
CAS Number:
Molecular Weight:
320.25

Products

Brand
Color
Feature
Formula Weight
Manufacturer
Physical Form
Product Line
Quality Segment
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing