Skip to Content
Merck
All Photos(1)

Key Documents

SAB1413125

Sigma-Aldrich

Monoclonal Anti-EGFR (phospho Y1068) antibody produced in rabbit

clone HOF-5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

antibody product type

primary antibodies

clone

HOF-5, monoclonal

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

flow cytometry: 1:40
immunofluorescence: 1:50-1:200
immunoprecipitation (IP): 1:100
western blot: 1:1000-1:2000

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr1068)

Gene Information

human ... EGFR(1956)

Immunogen

A synthetic phosphopeptide corresponding to residues surrounding Y1068 of human EGFR.

Physical form

In PBS, 150 mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service