Key Documents
S6644
Sigma-Aldrich
Sigma lab markers
Fine tip, blue
About This Item
UNSPSC Code:
41103032
NACRES:
NB.35
color
blue
General description
Writes on cold, wet surfaces. Quick-drying. Alcohol based permanent marker removed by alcohol.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Customers Also Viewed
Slide 1 of 1