Documents
Z743546
Teeny writer laboratory marker
twin-tip, fine / ultra fine, black
About This Item
UNSPSC Code:
41103032
NACRES:
NB.17
manufacturer/tradename
Diversified Biotech TNY-TWIN
color
black
Related Categories
General description
This Sharpie® pen has a fine tip at one end and an ultra fine tip at the other end. Recommended for writing on all our labels for samples that will be stored at room temperature and not exposed to chemicals and/or caustic agents.
Legal Information
Sharpie is a registered trademark of Sanford, LP