Key Documents
S5894
Sigma-Aldrich
Sigma lab markers
Fine tip, black
About This Item
UNSPSC Code:
41100000
Product S5894 is not currently sold in your country. Contact Technical Service
color
black
General description
Writes on cold, wet surfaces. Quick-drying. Alcohol based permanent marker removed by alcohol.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Customers Also Viewed
Slide 1 of 1