Skip to Content
Merck
All Photos(1)

Key Documents

S3144

Sigma-Aldrich

[D-Arg1, D-Phe5, D-Trp7,9, Leu11]-Substance P

≥95% (HPLC)

Synonym(s):

D-Arg-Pro-Lys-Pro-D-Phe-Gln-D-Trp-Phe-D-Trp-Leu-Leu-NH2, Antagonist D

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€174.00

€174.00


Please contact Customer Service for Availability


Select a Size

Change View
1 MG
€174.00

About This Item

Empirical Formula (Hill Notation):
C79H109N19O12
CAS Number:
Molecular Weight:
1516.83
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.32

€174.00


Please contact Customer Service for Availability

Quality Level

Assay

≥95% (HPLC)

storage temp.

−20°C

SMILES string

CC(C)C[C@H](NC(=O)[C@H](CC(C)C)NC(=O)[C@@H](Cc1c[nH]c2ccccc12)NC(=O)[C@H](Cc3ccccc3)NC(=O)[C@@H](Cc4c[nH]c5ccccc45)NC(=O)[C@H](CCC(N)=O)NC(=O)[C@@H](Cc6ccccc6)NC(=O)[C@@H]7CCCN7C(=O)[C@H](CCCCN)NC(=O)[C@@H]8CCCN8C(=O)[C@H](N)CCCNC(N)=N)C(N)=O

InChI

1S/C79H109N19O12/c1-46(2)38-59(68(83)100)91-70(102)60(39-47(3)4)92-73(105)64(43-51-45-88-56-28-14-12-25-53(51)56)95-72(104)61(40-48-20-7-5-8-21-48)93-74(106)63(42-50-44-87-55-27-13-11-24-52(50)55)94-69(101)57(32-33-67(82)99)89-71(103)62(41-49-22-9-6-10-23-49)96-76(108)66-31-19-37-98(66)78(110)58(29-15-16-34-80)90-75(107)65-30-18-36-97(65)77(109)54(81)26-17-35-86-79(84)85/h5-14,20-25,27-28,44-47,54,57-66,87-88H,15-19,26,29-43,80-81H2,1-4H3,(H2,82,99)(H2,83,100)(H,89,103)(H,90,107)(H,91,102)(H,92,105)(H,93,106)(H,94,101)(H,95,104)(H,96,108)(H4,84,85,86)/t54-,57+,58+,59+,60+,61+,62-,63-,64-,65+,66+/m1/s1

InChI key

XVOCEQLNJQGCQG-ACRSGXKRSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Arg-Pro-Lys-Pro-Phe-Gln-Trp-Phe-Trp-Leu-Leu-NH2

General description

The somatostatin gene codes for a peptide hormone containing 14 and 28 amino acid moieties. SST-14 is highly expressed in the central nervous system and peripheral organs. SST-28 is mainly expressed in enteroendocrine cells of the intestine.[1]

Biochem/physiol Actions

NK-1 tachykinin receptor antagonist
Somatostatin plays an important role in regulating the metabolism of other hormones such as growth hormone, insulin, IGF-1 (insulin-like growth factor 1), glucagon, glucagon-like peptide and gastrin. Somatostatin has both regulatory and inhibitory functions. It retards gastrointestinal motility and gallbladder contraction, suppresses gut exocrine secretion and reduces epithelial proliferation. Somatostatin is associated with intestinal nutrient absorption and blood flow regulation. This peptide hormone is believed to inhibit cell proliferation and promote apoptosis. Somatostatin acts as a neurotransmitter and neuromodulator, and is also associated in maintaining water balance.[1][2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael Patterson et al.
Peptides, 32(11), 2290-2294 (2011-08-13)
Ghrelin is a peptide hormone secreted into circulation from the stomach. It has been postulated to act as a signal of hunger. Ghrelin administration acutely increases energy intake in lean and obese humans and chronically induces weight gain and adiposity
J Cummings et al.
Journal of chromatography. B, Biomedical applications, 653(2), 195-203 (1994-03-04)
The neuropeptide growth factor antagonists [D-Arg1,D-Phe5,D-Trp7,9,Leu11]-substance P (D) and [Arg6,D-Trp7,9, [corrected] N-MePhe8]-substance P(6-11) (G) are currently undergoing preclinical evaluation as potential anticancer agents and clinical trials are planned for G in the near future. A reversed-phase high-performance liquid chromatographic separation
H Houben et al.
Peptides, 14(1), 109-115 (1993-01-01)
The effect of peptide and nonpeptide substance P antagonists on prolactin (PRL) and growth hormone (GH) secretion was evaluated in three-dimensional rat anterior pituitary cell aggregates. [D-Arg1,D-Phe5,D-Trp7,9,Leu11]Substance P inhibited basal growth hormone (GH) release at a concentration range of 1-10
S Langdon et al.
Cancer research, 52(16), 4554-4557 (1992-08-15)
The proliferation of small cell lung cancer (SCLC) cells appears sustained by multiple autocrine and paracrine circuits involving Ca2+ mobilizing neuropeptides. Consequently, broad spectrum neuropeptide antagonists which inhibit SCLC growth in vitro have been suggested as potential anticancer agents. Here
X H Gu et al.
Biochemical and biophysical research communications, 179(1), 130-133 (1991-08-30)
The specific binding of [125I]ET-1 to rat cardiac membrane fragments was inhibited by [D-Arg1,D-Phe, D-Try7,9,Leu11] substance P [substance P(D)], a potent bombesin antagonist. This inhibitory effect required high concentrations (greater than 3X10(-6)M) of substance P(D) and was accompanied by a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service