Skip to Content
Merck
All Photos(1)

Key Documents

Q0380

Sigma-Aldrich

Quinone Oxidoreductase 2 (NQO2) human

recombinant, expressed in E. coli, ≥90% (SDS-PAGE), ≥100 units/mg protein

Synonym(s):

NQO2 human, NRH nitroreductase human, NRH quinone reductase human

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

recombinant

expressed in E. coli

Quality Level

Assay

≥90% (SDS-PAGE)

form

lyophilized powder (one vial contains one mg protein)

specific activity

≥100 units/mg protein

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... NQO2(4835)

Application

Quinone oxidoreductase 2 has been used in a study to investigate whether recent advances in limiting paraquat′s toxicity makes it safer for use as an herbicide. Quinone oxidoreductase 2 has also been used in a study to investigate the NQO2-mediated toxicity of the chemotherapeutic drug CB1954.
The product from Sigma has been used to identify novel inhibitors of the enzyme NQO2.[1] It has also been used to investigate the effect of afobazole on quinone reductase 2 activity.[2]

Biochem/physiol Actions

Modulation of NQO2 activity can have effects on the levels and functions of the tumour suppressor, p53. Deletion of NQO2 can alter intra-cellular signalling via the NF-κB pathway.[1]
Unique human oxidoreductase that uses Dihydronicotinamide riboside (NRH) as a cosubstrate, not NAD(P)H. Activates anticancer prodrugs. Putative melatonin receptor.

Unit Definition

One unit will catalyze the oxidation of one micromole of 1-methyl-1,4-dihydronicotinamide to 1-methylnicotinamide per minute in the presence of menadione at pH 7.4 at 37 °C.

Physical form

contains 10% PBS buffer salts

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of Afobazole on Activity of Quinone Reductase 2
Kadnikov IA, et al.
Pharmaceutical Chemistry Journal, 47(10), 514-516 (2014)
Karen A Nolan et al.
Molecular cancer therapeutics, 11(1), 194-203 (2011-11-18)
The National Cancer Institute chemical database has been screened using in silico docking to identify novel nanomolar inhibitors of NRH:quinone oxidoreductase 2 (NQO2). The inhibitors identified from the screen exhibit a diverse range of scaffolds and the structure of one
Paraquat Research. Do Recent Advances In Limiting Its Toxicity Make Its Use Safer?
Baltazar, T., et al.
British Journal of Pharmacology (2012)
Faisal Hayat et al.
International journal of molecular sciences, 22(3) (2021-01-28)
As catabolites of nicotinamide possess physiological relevance, pyridones are often included in metabolomics measurements and associated with pathological outcomes in acute kidney injury (AKI). Pyridones are oxidation products of nicotinamide, its methylated form, and its ribosylated form. While they are
Qiuying Cheng et al.
Biochemical pharmacology, 83(1), 139-148 (2011-10-18)
Carbonyl reductase activity catalyzes the two electron reduction of several endogenous and exogenous carbonyl substrates. Recent data indicate that the expression of human carbonyl reductase 3 (CBR3) is regulated by the master redox switch Nrf2. Nrf2 binds to conserved antioxidant

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service