Skip to Content
Merck
All Photos(1)

Key Documents

SRP6539

Sigma-Aldrich

NQO1 human

recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Synonym(s):

Azoreductase, DT-Diaphorase (DTD), Dioxin-inducible 1, Disphorase-4 (DIA4), Menadione Reductase, NAD(P)H Dehydrogenase (Quinine) 1 (NQO1), NAD(P)H Quinone Oxireductase, Phylloquinone Reductase, Quinone Oxidoreductase 1, Quinone Reductase 1 (QR1)

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€411.00

€411.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
€411.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.77

€411.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

human

recombinant

expressed in E. coli

Assay

≥90% (SDS-PAGE)

form

liquid

mol wt

~30 kDa (Human NQO1 aa 1-274 is fused at the C-terminus to a His-tag.)

packaging

pkg of 10 and 50 μg

impurities

<0.1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... NQO1(1728)

General description

Research area: Cell signaling. NQO1 (NAD(P)H quinone dehydrogenase 1) is an obligate two-electron reductase. It is a cytoplasmic, homodimeric protein, with one FAD molecule per monomer. This gene is localized to human chromosome 16q23.[1][2] It shows a wide range of tissue expression.[3]

Biochem/physiol Actions

NQO1 (NAD(P)H quinone dehydrogenase 1) uses NADH or NADPH as a reducing cofactor, and is characterized by its inhibition by dicoumarol. It is thought to confer protection against natural and exogenous quinones.[1] It is involved in the conversion of benzene derived quinones to less toxic hydroquinones, and participates in benzene-associated hematotoxicity. It is involved in the reduction and eventual activation of few chemotherapeutic drugs and of environmental carcinogens, such as heterocyclic amines, nitroaromatic compounds, and possibly cigarette smoke particulate.[2] C609T null variant of this gene is linked with breast cancer susceptibility in north Indian patients.[4] NQO1 is up-regulated in serous ovarian carcinoma, and this expression is linked with poor prognosis; hence, this gene has potential as biomarker for the same.[5]

Physical form

0.2 μm-filtered solution in 50 mM Tris-Cl, pH 8.0, containing 1 mM DTT.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

NAD(P)H:quinone acceptor oxidoreductase 1 (NQO1), a multifunctional antioxidant enzyme and exceptionally versatile cytoprotector.
Dinkova-Kostova AT and Talalay P.
Archives of Biochemistry and Biophysics, 501(1), 116-123 (2010)
Prevalence of the inactivating 609C-->T polymorphism in the NAD(P)H:quinone oxidoreductase (NQO1) gene in patients with primary and therapy-related myeloid leukemia.
Larson RA, et al.
Blood, 94(2), 803-807 (1999)
High expression of NQO1 is associated with poor prognosis in serous ovarian carcinoma.
Cui X, et al.
BMC Cancer, 15:244 (2015)
The C609T (Pro187Ser) Null Polymorphism of the NQO1 Gene Contributes Significantly to Breast Cancer Susceptibility in North Indian Populations: a Case Control Study.
Yadav P, et al.
Asian Pacific Journal of Cancer Prevention, 17(3), 1215-1219 (2016)
Xiang Li et al.
Bioorganic & medicinal chemistry, 24(5), 1006-1013 (2016-01-25)
A new series of ortho-naphthoquinone analogs of β-lapachone were designed, synthesized and evaluated. The biological results indicated that most of our compounds were efficient substrates for NQO1. The new scaffold with water-soluble side chain resulted in greater solubility under acidic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service