Skip to Content
Merck
All Photos(1)

Key Documents

P8874

Sigma-Aldrich

Monoclonal Anti-Phosphacan antibody produced in mouse

~2 mg/mL, clone 122.2, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-PTPRB, Anti-Receptor-type Protein-tyrosine phosphatase β

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
€624.00

€624.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
200 μL
€624.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

€624.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

122.2, monoclonal

form

buffered aqueous solution

mol wt

antigen ~180 kDa (higher band may be present)

species reactivity

rat

packaging

antibody small pack of 25 μL

concentration

~2 mg/mL

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: 0.2-0.4 μg/mL using total extract of rat brain

isotype

IgM

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Monoclonal Anti-Phosphacan (mouse IgM isotype) is derived from the hybridoma 122.2 produced by the fusion of mouse myeloma cells (P3X cells) and splenocytes from BALB/c mice immunized with rat brain proteoglycans. Chondroitin sulfate proteoglycans are neural cell adhesion molecules (NCAM) ligands present in the brain extracellular matrix (ECM). Phosphacan protein is expressed mainly in astrocytes and is a ligand for NCAM. Phosphacan is the soluble extracellular domain of the receptor-type transmembrane protein tyrosine phosphatase (RPTPb).

Immunogen

rat brain proteoglycans.

Application

Monoclonal Anti-Phosphacan antibody produced in mouse has been used in:
  • immunoblotting
  • immunohistochemistry
  • immunocytochemistry.

Biochem/physiol Actions

Phosphacan levels elevates during late embryogenesis. It reaches a plateau two weeks postnatal before reaching stable. Receptor-type transmembrane protein tyrosine phosphatase (RPTPb) functions to promote primary tecal neurons neurite growth, neural migration and also induces cell adhesion. Both phosphacan and RPTPb can bind to NCAM and tenascin-C and −R. Phosphacan can oppose RPTPb by competing for its binding sites. Both in hippocampal and spinal cord neurons, phosphacan can affect neuronal adhesion and neurite outgrowth.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

RGMa mediates reactive astrogliosis and glial scar formation through TGFbeta1/Smad2/3 signaling after stroke
Zhang R, et al.
Cell Death and Differentiation, 25(8), 1503-1503 (2018)
Receptor protein tyrosine phosphatases in nervous system development
Johnson KG and Van Vactor D
Physiological Reviews, 83(1), 1-24 (2003)
The tissue plasminogen activator (tPA/Plasmin) extracellular proteolytic system regulates seizure-induced hippocampal mossy fiber outgrowth through a proteoglycan substrate
Wu YP, et al.
The Journal of cell biology, 148(6), 1295-1304 (2000)
Yuan Qin et al.
Acta pharmacologica Sinica, 40(6), 724-736 (2018-10-14)
Increasing evidence suggests that Ras-related in brain 7 (Rab7), an endosome-localized small GTPase contributes to cerebral ischemic brain injury. In the present study, we investigated the role of Rab7 in ischemic stroke-induced formation of astrogliosis and glial scar. Rats were
Nuttapong Yawoot et al.
Journal of cellular physiology, 237(3), 1818-1832 (2021-11-27)
Even though astrocytes have been widely reported to support several brain functions, studies have emerged that they exert deleterious effects on the brain after ischemia and reperfusion (I/R) injury. The present study investigated the neuroprotective effects of melatonin on the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service