Skip to Content
Merck
All Photos(1)

Key Documents

P169

Supelco

Protriptyline hydrochloride

Synonym(s):

N-Methyl-5H-dibenzo[a,d]cycloheptene-5-propanamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H21N · HCl
CAS Number:
Molecular Weight:
299.84
EC Number:
MDL number:
UNSPSC Code:
41116107

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

SMILES string

Cl[H].CNCCCC1c2ccccc2C=Cc3ccccc13

InChI

1S/C19H21N.ClH/c1-20-14-6-11-19-17-9-4-2-7-15(17)12-13-16-8-3-5-10-18(16)19;/h2-5,7-10,12-13,19-20H,6,11,14H2,1H3;1H

InChI key

OGQDIIKRQRZXJH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

replaced by

Product No.
Description
Pricing

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nonsurgical management of the obstructive sleep apnea patient.
W K Thornton et al.
Journal of oral and maxillofacial surgery : official journal of the American Association of Oral and Maxillofacial Surgeons, 54(9), 1103-1108 (1996-09-01)
K E Kinnamon et al.
Proceedings of the Society for Experimental Biology and Medicine. Society for Experimental Biology and Medicine (New York, N.Y.), 216(3), 424-428 (1997-12-24)
Forty-nine "standard" compounds known to be useful in the treatment of other diseases were tested for their suppressive activity against the trypomastigotes of Trypanosoma cruzi-infected mice. The most active was the antidepressant protriptyline, which was almost three times as effective
Y Huang
British journal of pharmacology, 117(3), 533-539 (1996-02-01)
1 The effects of noradrenaline (NA) uptake inhibitors on contractions induced by NA, high K+, and 12-O-tetradecanoylphorbol-13-acetate (TPA) in rat isolated aorta were investigated. 2 Protriptyline (0.3 microM) and amitriptyline (0.3 microM) produced an approximately parallel shift to the right
J R Wooltorton et al.
British journal of pharmacology, 116(4), 2191-2200 (1995-10-01)
1. The block of K+ currents by amitriptyline and the related tricyclic compounds cyproheptadine and dizocilpine was studied in dissociated rat sympathetic neurones by whole-cell voltage-clamp recording. 2. Cyproheptadine (30 microM) inhibited the delayed-rectifier current (Kv) by 92% and the
B T Duhart et al.
Xenobiotica; the fate of foreign compounds in biological systems, 29(7), 733-746 (1999-08-24)
1. The potential of various fungi to metabolize protriptyline (an extensively used antidepressant) was studied to investigate similarities between mammalian and microbial metabolism. 2. Metabolites produced by each organism were isolated by high-pressure liquid chromatography and identified by nuclear magnetic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service