Skip to Content
Merck
All Photos(1)

Key Documents

MAS010

Sigma-Aldrich

Oxalate Quick Test Strips

Sufficient for 10 tests

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€76.80

€76.80


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 KIT
€76.80

About This Item

UNSPSC Code:
12352106
NACRES:
NA.84

€76.80


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

input

serum
plasma
urine

application(s)

food and beverages

detection method

colorimetric

relevant disease(s)

genitourinary disorders

storage temp.

2-8°C

General description

Oxalate, or Oxalic Acid, is a metabolic breakdown product of the Kreb′s Cycle in eukaryotes and the glyoxylate cycle in other microorganisms.Typically present in the urine of humans and other mammals, oxalate concentration can be used as a measure of kidney function. A high level of oxalate is an indicator for kidney stones, which are primarily made of the insoluble salt calcium oxalate. Measuring oxalate is more accurate than measuring calcium as a marker for kidney stones because calcium is excreted at high concentrations even in normal urine.

Application

The Oxalate Quick Test Strips are designed for determining oxalate levels in the following applications:
  • Food and Beverage Testing
  • Biological samples such as plasma, serum, and urine.

Features and Benefits

  • Rapid and Sensitive: Provides swift results with only 10 μL of sample for semi-quantitative measurement within a range of 0-1.5 mg/dL or 0-170 μM (undiluted) Oxalate, ensuring quick and accurate analysis.
  • Efficient: Sample treatment and assay completion take less than 10 minutes, saving time and enhancing productivity.
  • User-Friendly: Convenient test strip design eliminates the need for a plate or cuvette reader, simplifying the testing process and making it accessible for users of all skill levels.

Suitability

Suitable for the detection of oxalate concentrations in serum, plasma, urine, fruits or vegetables, etc.

Principle

Oxalate Quick Test Strips are based on the enzymatic oxidation of oxalate coupled with a color reaction in one step. The intensity of the product color is directly proportional to the oxalate concentration in the sample.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–3 of 3 Questions  
  1. 1.Provide a sample preparation method that is specific to vegetable powders and specify the required reagents?
    2.What amount of sample should be used and what volume of water or sample-to-water ratio is recommended for roughly ground dried leaves, equivalent to spinach?
    3.Following the homogenization of the roughly ground sample in water, should I centrifuge the mixture before using the test strip, and if so, at what speed and for how long? Alternatively, can the mixture be used as is?

    1 answer
    1. 1.The MAS010 strips have been tested with both fresh and dried leaves such as spinach and green tea. The leaves were homogenized with distilled water using a Dounce homogenizer and diluted as needed to ensure that the oxalate concentration falls within the detection range of the strip (0.25-1.5 mg/dL). Therefore, the sample preparation for vegetable powder should be similar to that for leaves. It is recommended to dissolve the vegetable powder in water and test the sample to ensure that the oxalate concentration falls within the detection range. If the concentration is too high, perform dilutions to find the appropriate dilution factor for the strips. Conversely, if the concentration is too low, dissolve a greater amount of vegetable powder in the same volume of water. When testing different dilution factors, the strips can be conserved by cutting them in half, but remember to use the full-sized strip for the final test. For further inquiries, feel free to reach out.
      2.Spinach has a high concentration of oxalate (1029 mg soluble oxalate/100 g leaves). For a sample similar to spinach, try homogenizing 5 mg of the dried leaves with 10 mL of water, aiming for a concentration of 0.5 mg/dL oxalate, the mid-range of the detection of the strips. If measuring the total oxalate content, homogenize the sample in 2M HCl instead of water. Spinach has about 1959 mg total oxalate/100 g leaves. For a concentration of 0.5 mg/dL oxalate, mix 5 mg of dried leaves with 10 mL of 2M HCl, then neutralize the sample with 10 mL of 2M NaOH before transferring it onto the strip.
      3.A Dounce homogenizer or a mortar and pestle can be used to further mash the leaves. Subsequently, the sample can be centrifuged at 10,000 g for 5 minutes, and the clear supernatant can be transferred to the strips. The speed and time of centrifugation do not matter as long as the particulates can be pelleted at the bottom.

      Helpful?

  2. Can this test oxalate levels in cooked foods? Is there an indicator of the quantitative amount of oxalates?

    1 answer
    1. Unfortunately, the kit MAS010 hasn't been tested for cooked foods but most likely it can work. The kit contains an oxalate chart that works as the indicator of quantitative amount of oxalates.

      Helpful?

  3. I'm looking for a way to do a quick/qualitative at home screening for oxalates. Will this product be useful for that? If so, what is needed other than the strips?

    1 answer
    1. Unless otherwise stated this product is intended for research use only and is not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses, or any type of consumption or application to humans or animals.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service