Skip to Content
Merck
All Photos(1)

Key Documents

MAK502

Sigma-Aldrich

Galactose Assay Kit

sufficient for 100 colorimetric or fluorometric tests

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€472.00

€472.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 KIT
€472.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

€472.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

application(s)

cosmetics
food and beverages
pharmaceutical

detection method

colorimetric
fluorometric

relevant disease(s)

pediatric diseases; neonatal diseases

shipped in

wet ice

storage temp.

−20°C

General description

Galactose (C6H12O6) is a monosaccharide that is found in dairy products, sugar beets, gums and mucilages. It is also synthesized in mammals, where it forms part of glycolipids and glycoproteins in several tissues. It forms the disaccharide lactose when combined with glucose.

Features and Benefits

  • Linear detection range: 10 to 1000 µM galactose for the colorimetric assay and 10 to 100 µM for the fluorometric assay.

Suitability

The kit is suitable for determining galactose concentrations in serum, plasma, urine, saliva, milk, culture medium, food and beverage products, and other biological samples, as well as for studying the effects of drugs on galactose metabolism.

Principle

The Galactose Assay Kit uses specific enzyme-coupled reactions to form a colored product. The color intensity at 570 nm or fluorescence intensity at λEx = 530 nm/λEm = 585 nm is directly proportional to the galactose concentration in the sample.

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service