Skip to Content
Merck
All Photos(1)

Key Documents

MAK487

Sigma-Aldrich

Lactose Assay Kit

sufficient for 100 colorimetric or fluorometric tests

Synonym(s):

Lactose Detection Kit

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€427.00

€427.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 KIT
€427.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

€427.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

application(s)

agriculture
food and beverages
pharmaceutical

detection method

colorimetric
fluorometric

relevant disease(s)

gastrointestinal diseases

shipped in

wet ice

storage temp.

−20°C

General description

Lactose (C12H22O11) is a disaccharide comprised of galactose and glucose which form a β1-4 glycosidic linkage. Lactose is the major sugar in milk, making up around 2–8 % of milk (by weight).

Features and Benefits

  • Linear detection range of the kit is 17 to 2000 µM lactose for the colorimetric assay and 6 to 100 µM for the fluorometric assay.

Suitability

The kit is suitable for lactose activity determination in milk, food, beverages and other biological samples, as well as for studying the effects of drugs on lactose metabolism.

Principle

The Lactose Assay Kit uses a specific enzyme-coupled reaction in which lactose is cleaved and the resulting galactose forms a colored product. The color intensity at 570 nm or fluorescence intensity at λEx = 530 nm/λEm = 585 nm is directly proportional to the lactose concentration in the sample.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service