Skip to Content
Merck
All Photos(1)

Key Documents

M8128

Sigma-Aldrich

Maltopentaose

≥95%

Synonym(s):

Maltopentose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H52O26
CAS Number:
Molecular Weight:
828.72
Beilstein:
79498
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

biological source

corn starch (plant)

Assay

≥95%

form

powder

SMILES string

OC[C@H]1O[C@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](O[C@@H]2CO)O[C@H]3[C@H](O)[C@@H](O)[C@H](O[C@@H]3CO)O[C@H]4[C@H](O)[C@@H](O)[C@H](O[C@@H]4CO)O[C@H]5[C@H](O)[C@@H](O)[C@@H](O)O[C@@H]5CO)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C30H52O26/c31-1-6-11(36)12(37)18(43)27(49-6)54-23-8(3-33)51-29(20(45)14(23)39)56-25-10(5-35)52-30(21(46)16(25)41)55-24-9(4-34)50-28(19(44)15(24)40)53-22-7(2-32)48-26(47)17(42)13(22)38/h6-47H,1-5H2/t6-,7-,8-,9-,10-,11-,12+,13-,14-,15-,16-,17-,18-,19-,20-,21-,22-,23-,24-,25-,26+,27-,28-,29-,30-/m1/s1

InChI key

FTNIPWXXIGNQQF-DWTFCAFKSA-N

Looking for similar products? Visit Product Comparison Guide

Application

A partial hydrolysis of maltopentaose has been used to prepare a dextran ladder in a study that assessed cationic labeling of oligosaccharides. Maltopentaose has also been used in a study to investigate glycation and phosphorylation of α-lactalbumin.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anja Warmerdam et al.
SpringerPlus, 2, 402-402 (2013-09-12)
Enzymatic synthesis of galacto-oligosaccharides is usually performed at high initial substrate concentrations since higher yields are obtained. We report here on the stability of β-galactosidase from Bacillus circulans at 25, 40, and 60°C in buffer, and in systems with initially
Jakub Ujma et al.
Journal of the American Society for Mass Spectrometry, 30(6), 1028-1037 (2019-04-13)
There is increasing biopharmaceutical interest in oligosaccharides and glycosylation. A key requirement for these sample types is the ability to characterize the chain length, branching, type of monomers, and importantly stereochemistry and anomeric configuration. Herein, we showcase the multi-function capability
Toshiaki Kozuka et al.
Plant physiology, 182(2), 1114-1129 (2019-11-22)
Exposure of dark-grown (etiolated) seedlings to light induces the heterotrophic-to-photoautotrophic transition (de-etiolation) processes, including the formation of photosynthetic machinery in the chloroplast and cotyledon expansion. Phytochrome is a red (R)/far-red (FR) light photoreceptor that is involved in the various aspects
Min Ma et al.
Journal of agricultural and food chemistry, 67(12), 3380-3388 (2019-02-27)
α-Glucosidase from Aspergillus niger (AgdA; typical α-1,4-glucosidase) is known to industrially produce α-(1→6)-glucooligosaccharides. This fungus also has another α-glucosidase-like protein, AgdB. To learn its function, wild-type AgdB was expressed in Pichia pastoris. However, the enzyme displayed two electrophoretic forms due
H Enomoto et al.
Journal of dairy science, 92(7), 3057-3068 (2009-06-17)
Alpha-lactalbumin (alpha-LA) was glycated with maltopentaose (MP) through the Maillard reaction (MP-alpha-LA) and subsequently phosphorylated by dry heating in the presence of pyrophosphate to investigate its structure and physiological functions. Glycation occurred effectively, and the sugar content of alpha-LA increased

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service