Skip to Content
Merck
All Photos(1)

Key Documents

SMB01321

Sigma-Aldrich

Maltopentaose

95% (HPLC)

Synonym(s):

Amylopentaose, Maltopentose

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€83.30
50 MG
€334.00

€83.30


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€83.30
50 MG
€334.00

About This Item

Empirical Formula (Hill Notation):
C30H52O26
CAS Number:
Molecular Weight:
828.72
Beilstein:
79498
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

€83.30


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

95% (HPLC)

form

solid

color

white

solubility

water: 50 g/L

shipped in

ambient

SMILES string

OC[C@H]1O[C@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](O[C@@H]2CO)O[C@H]3[C@H](O)[C@@H](O)[C@H](O[C@@H]3CO)O[C@H]4[C@H](O)[C@@H](O)[C@H](O[C@@H]4CO)O[C@H]5[C@H](O)[C@@H](O)[C@@H](O)O[C@@H]5CO)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C30H52O26/c31-1-6-11(36)12(37)18(43)27(49-6)54-23-8(3-33)51-29(20(45)14(23)39)56-25-10(5-35)52-30(21(46)16(25)41)55-24-9(4-34)50-28(19(44)15(24)40)53-22-7(2-32)48-26(47)17(42)13(22)38/h6-47H,1-5H2/t6-,7-,8-,9-,10-,11-,12+,13-,14-,15-,16-,17-,18-,19-,20-,21-,22-,23-,24-,25-,26+,27-,28-,29-,30-/m1/s1

InChI key

FTNIPWXXIGNQQF-DWTFCAFKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Maltopentaose, also known as Amylopentaose, is a naturally occurring oligosaccharide and endogenous metabolite composed of five α-1,4-linked glucose molecules. It is the shortest chain pentasaccharide categorized under maltodextrin and finds applications in exploring glycation and phosphorylation of α-lactalbumin. Notably, at a concentration of 1 mM, maltopentaose enhances the α-amylase synthesis rate in B. stearothermophilus. Additionally, it serves as a substrate for porcine pancreatic α-amylase (PPA), making it valuable for investigating various PPA inhibitors.

Application

A partial hydrolysis of maltopentaose has been used to prepare a dextran ladder in a study that assessed cationic labeling of oligosaccharides.
Maltopentaose is a versatile compound that finds application in metabolomics and biochemical research.

Features and Benefits

  • High-purity compound suitable for a wide variety of research applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.
To gain a comprehensive understanding of our extensive range of Oligosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Enomoto et al.
Journal of dairy science, 92(7), 3057-3068 (2009-06-17)
Alpha-lactalbumin (alpha-LA) was glycated with maltopentaose (MP) through the Maillard reaction (MP-alpha-LA) and subsequently phosphorylated by dry heating in the presence of pyrophosphate to investigate its structure and physiological functions. Glycation occurred effectively, and the sugar content of alpha-LA increased
Roberto Quezada-Calvillo et al.
The Journal of nutrition, 138(4), 685-692 (2008-03-22)
The detailed mechanistic aspects for the final starch digestion process leading to effective alpha-glucogenesis by the 2 mucosal alpha-glucosidases, human sucrase-isomaltase complex (SI) and human maltase-glucoamylase (MGAM), are poorly understood. This is due to the structural complexity and vast variety

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service