All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H11N3O5
Molecular Weight:
205.17
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
Recommended Products
Assay
≥90% (TLC)
form
solid
shipped in
wet ice
storage temp.
−20°C
SMILES string
OC[C@H]1O[C@H](N=[N+]=[N-])[C@@H](O)[C@@H](O)[C@@H]1O
InChI
1S/C6H11N3O5/c7-9-8-6-5(13)4(12)3(11)2(1-10)14-6/h2-6,10-13H,1H2/t2-,3-,4+,5+,6+/m1/s1
InChI key
KSRDTSABQYNYMP-PQMKYFCFSA-N
General description
α-D-Mannopyranosyl is a glycosyl azide.
Application
α-D-Mannopyranosyl azide has been used in a study to assess the photochemistry of glycosyl azides. It has also been used in a study to investigate stereoselective photochemical transformations of hexopyranosyl imides.
Used to make N-link glycopeptides in glycobiology.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Stereoselective photochemical transformations of hexopyranosyl imides to highly functionalised heterocycles
Thiering, S., et al.
Journal of the Chemical Society. Perkin Transactions 1, 8, 801-806 (2001)
Photochemistry of glycosyl azides-II1: Investigation of the dual behavior: formation of a reversible intermediate and chain-degradation
Szarek, W., et al.
Tetrahedron, 34, 1427-1433 (1978)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service