Skip to Content
Merck
All Photos(1)

Key Documents

M024080

Sigma-Aldrich

ssH-Linker

configured for PerkinElmer, configured for Polygen

Synonym(s):

9,11-Dioxa-2-aza-10-phosphatridecanoic acid, 10-[bis(1-methylethyl)amino]-13-cyano-, 2-[[(4-methoxyphenyl)diphenylmethyl]amino]ethyl este

Sign Into View Organizational & Contract Pricing

Select a Size

0.25 G
€151.00
1 G
€512.00

€151.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
0.25 G
€151.00
1 G
€512.00

About This Item

Empirical Formula (Hill Notation):
C38H53N4O5P
Molecular Weight:
676.83
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.51

€151.00


Please contact Customer Service for Availability

Request a Bulk Order

type

for DNA synthesis

product line

Proligo Reagents

Assay

≥98% (31P-NMR)
≥98.0% (reversed phase HPLC)

compatibility

configured for PerkinElmer
configured for Polygen

storage temp.

−20°C

SMILES string

COc1ccc(cc1)C(NCCOC(=O)NCCCCCCOP(OCCC#N)N(C(C)C)C(C)C)(c2ccccc2)c3ccccc3

InChI

1S/C38H53N4O5P/c1-31(2)42(32(3)4)48(47-29-16-25-39)46-28-15-7-6-14-26-40-37(43)45-30-27-41-38(33-17-10-8-11-18-33,34-19-12-9-13-20-34)35-21-23-36(44-5)24-22-35/h8-13,17-24,31-32,41H,6-7,14-16,26-30H2,1-5H3,(H,40,43)

InChI key

VTFLXOVDKJUHER-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

ssH-linker comprises an internal carbamate group attached to the MMT-protected amino group via a short spacer. The carbamate moiety facilitates the cleavage of the MMT group under mildly acidic conditions while increasing the stability of the MMT group during the deprotection of the oligonucleotide with ammonia. The carbamate group also enhances the reactivity of the amino group through a neighbor group effect, and thereby accelerates conjugations to amino reactive modifiers and reporters. The MMT-group of ssH-linker serves as an excellent purification handle after the oligonucleotide synthesis in trityl-on mode, similar to the DMT-group of conventional oligonucleotides. The MMT-group is cleaved under very mild conditions in aqueous acetic acid (10% glacial acetic acid in the water, 20 min. room temp.) Alternatively, the MMT-group can be cleaved on the synthesis instrument with an acidic deblock solution to enable on-support labeling protocols.

Legal Information

This product is covered under US Patent Number 7,491,857

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service