
ssH-Linker

Synonym(s):
9,11-Dioxa-2-aza-10-phosphatridecanoic acid, 10-[bis(1-methylethyl)amino]-13-cyano-, 2-[[(4-methoxyphenyl)diphenylmethyl]amino]ethyl este
Empirical Formula (Hill Notation):
C38H53N4O5P
Molecular Weight:
676.83
Products
Brand
Formula Weight
Product Line
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing