Skip to Content
Merck
All Photos(1)

Key Documents

L9895

Sigma-Aldrich

Lectin from Pseudomonas aeruginosa

lyophilized powder

Synonym(s):

PA-I

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€261.00
2 MG
€446.00

€261.00


Please contact Customer Service for Availability


Select a Size

Change View
1 MG
€261.00
2 MG
€446.00

About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

€261.00


Please contact Customer Service for Availability

form

lyophilized powder

Quality Level

potency

<15 μg/mL per mL agglutination activity

storage temp.

−20°C

Biochem/physiol Actions

PA-I has a major affinity for D-galactose and its derivatives. PA-I agglutinates all types of human and animal erythrocytes and interacts also with unicellular protozoa, algae and bacteria. PA-I induces mitogenic stimulation in cultured human lymphocytes pretreated with neuraminidase.

Packaging

Package size based on protein content. Vial content based on protein by biuret.

Quality

Highly purified.

Physical form

Lyophilized powder containing approx. 80% protein (biuret), balance primarily sodium citrate, plus traces of calcium chloride, magnesium chloride and manganese chloride

Preparation Note

Purified by affinity chromatography using a modification of Gilboa-Garber.1

Analysis Note

Agglutination activity is expressed in μg/ml and is determined from serial dilutions of a 1 mg/ml solution using phosphate buffered saline, pH 7.0. This activity is the lowest concentration to agglutinate a 2% suspension of neuraminidase treated human blood group A erythrocytes after one hr incubation at 25 °C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alessandro Palmioli et al.
Chembiochem : a European journal of chemical biology, 20(23), 2911-2915 (2019-06-20)
Biofilm formation by bacterial pathogens is a hallmark of chronic infections and is associated to increased antibiotic tolerance that makes pathogens difficult to eradicate with conventional antibiotic therapies. Infections caused by Pseudomonas aeruginosa are of great concern, especially for immunocompromised
Akiko Chiba et al.
Molecular cancer research : MCR, 18(1), 130-139 (2019-10-20)
Breast tumors have their own specific microbiota, distinct from normal mammary gland tissue. Patients with breast cancer that present with locally advanced disease often undergo neoadjuvant chemotherapy to reduce tumor size prior to surgery to allow breast conservation or limit

Articles

Find the right lectin for your research with our lectin selection guide, organized by lectin source/species, carbohydrate specificity, blood group specificity, and more.

Questions

  1. Which one is this, LecA or LecB?

    Which one is this, LecA or LecB?

    1 answer
    1. Based on the production protocol of this product, it is directed towards the purification of PA-I only, with PA-II removal. According to the literature, PA-I is LecA and PA-II is LecB. Therefore, it appears that this product is LecA. For more information, please refer to the following literature: [Literature Link](https://www.ncbi.nlm.nih.gov/pmc/articles/PMC1698087/)

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service