MBD0016
Inactivated Pseudomonas aeruginosa
Suitable for DNA extraction, PCR, sequencing, next generation sequencing, >10^8 bacteria/ml
Select a Size
€248.00
Select a Size
About This Item
€248.00
Recommended Products
Quality Level
form
liquid
concentration
>10^8 bacteria/ml
technique(s)
DNA extraction: suitable
DNA sequencing: suitable
PCR: suitable
shipped in
dry ice
storage temp.
−20°C
Related Categories
General description
Pseudomonas aeruginosa is a gram negative aerobic to facultative anaerobe rod shaped bacteria which commonly inhabits soil and aqueous environments. In cases of a compromised immune system Pseudomonas aeruginosa can be an opportunistic pathogen. Its devastating effect is mostly known in Cystic Fibrosis (CF) patients. In addition, it often poses a major problem in burn wounds, chronic wounds, and in chronic obstructive pulmonary Disorder (COPD). P.aeruginosa forms biofilms on implanted biomaterials within hospital surface and water supplies. The international nosocomial infection control consortium reported that P. aeruginosa nosocomial infections have become a worldwide healthcare issue.
Read here how to use our standards to ensure data integrity for your microbiome research.
Application
Suitable for Quantitative standard for PCR, Sequencing and NGS
Features and Benefits
- Individual microbial standard for microbiomics and meta-genomics workflow
- Suitable standard for PCR, sequencing and NGS
- Improve Bioinformatics analyses
- Increases reproducibility
- Compare results lab to lab
Physical form
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
DNA standards enhance metagenomics research integrity, offering precise species study and mixed community standards.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service