Skip to Content
Merck
All Photos(2)

Key Documents

L0657

Sigma-Aldrich

Lard oil

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
€102.00
1 L
€175.00

€102.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
500 ML
€102.00
1 L
€175.00

About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€102.00


Please contact Customer Service for Availability

Request a Bulk Order

form

liquid

Quality Level

impurities

~0.5% free fatty acids

functional group

oleic acid

lipid type

oils

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Lard oil may be used to test the properties of antioxidants such as flavonoids and as a producer of mutagens and oxidants in oil smoke.

Storage Class Code

10 - Combustible liquids

WGK

awg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P F Wu et al.
Mutation research, 403(1-2), 29-34 (1998-09-03)
According to earlier studies, fumes from cooking oils were found to be mutagenic and several polycyclic aromatic hydrocarbons (PAHs), (benzo(a)pyrene (B(a)P), benz(a)antracene (B(a)A), and dibenz(a,h)anthracene (DB(ah)A)) were identified. Fume samples from three different commercial cooking oils frequently used in Taiwan
Li Ping-Chia et al.
Journal of biomedical science, 16, 58-58 (2009-07-07)
Airborne particulate matter, from cooking oil, smoking, engine exhaust and other sources, is associated with the development of atherosclerosis and myocardial infarction. In order to explore the cellular and molecular events following exposure of rats to lard oil smoke, we
T A Chiang et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 37(2-3), 125-134 (1999-05-05)
According to toxicological studies, there are several unidentified mutagens derived from cooking oil fumes appearing in kitchens of Chinese homes where women daily prepare food. Data are limited to an analysis of aromatic amines from cooking oil fumes, which are
Kai-Wen Huang et al.
Molecular therapy. Nucleic acids, 19, 361-370 (2019-12-27)
Non-alcoholic fatty liver disease (NAFLD) culminates in insulin resistance and metabolic syndrome. Because there are no approved pharmacological treatment agents for non-alcoholic steatohepatitis (NASH) and NAFLD, different signaling pathways are under investigation for drug development with the focus on metabolic
Liangping Zhao et al.
Gene, 771, 145354-145354 (2020-12-18)
Puerarin shows inhibitory effects on inflammation in chronic heart failure (CHF), but its efficacy in coronary heart disease (CHD) remained vague. Rat CHD model was constructed, and serum parameters were determined using a blood liquid biochemical analyzer. Also, contents of

Questions

  1. What is the known total lipid content of this analytical lard oil standard? Or is there any documentation on fatty acid or triglyceride content for this substance?

    1 answer
    1. This product is not tested for use as an analytical standard. This material is of porcine origin, however, the total lipid content is not determined. A previous porcine lard oil product, which is no longer available, was primarily made up of oleic acid (53 %), palmitic acid (21 %), linoleic acid (10 %), and steric acid (4-5 %). This product may have a similar profile. The free fatty acids content is listed on the lot-specific certificate of analysis.

      Please see the link below to review a sample Certificate:
      https://www.sigmaaldrich.com/certificates/sapfs/PROD/sap/certificate_pdfs/COFA/Q14/L0657-BULKSLCR5893.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service