Skip to Content
Merck
All Photos(1)

Key Documents

K0145

Sigma-Aldrich

KALA Amphipathic Peptide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H-Trp-Glu-Ala-Lys-Leu-Ala-Lys-Ala-Leu-Ala-Lys-Ala-Leu-Ala-Lys-His-Leu-Ala-Lys-Ala-Leu-Ala-Lys-Ala-Leu-Lys-Ala-Cys-Glu-Ala-OH
Molecular Weight:
3131.82
UNSPSC Code:
12352202

form

lyophilized solid

Quality Level

shipped in

wet ice

storage temp.

−20°C

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suan P Quah et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 108(3), 834-844 (2019-06-25)
We report the structural and mechanical behavior of multicomponent hydrogels comprising the commercial poly(ethylene oxide)-poly(propylene oxide)-poly(ethylene oxide) block copolymer F127 and alginate. Previous studies on this system have shown thermoreversible behavior in shear rheology. Here we explore the properties of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service