Skip to Content
Merck
All Photos(6)

Key Documents

HPA038568

Sigma-Aldrich

Anti-MADD antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Denn, Anti-Kiaa0358, Anti-Map-kinase activating death domain, Anti-Rab3gep

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€540.00

€540.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€540.00

About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€540.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:1000-1:2500

immunogen sequence

YSQQINEVLDQLANLNGRDLSIWSSGSRHMKKQTFVVHAGTDTNGDIFFMEVCDDCVVLRSNIGTVYERWWYEKLINMTYCPKTKVLCLWRRNGSETQ

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MADD(8567)

General description

MADD (MAP kinase-activating death domain protein) is located on human chromosome 11p11 and it consists of 36 exons. It is also called as DENN or Rab3GEP. The IG20 (insulinoma glucagonoma clone 20) gene codes for MADD.

Immunogen

MAP-kinase activating death domain recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

MADD (MAP kinase-activating death domain protein) prevents apoptosis and induces human lung adenocarcinoma. It has the ability to bind to the death domain of TNFR1 (tumor necrosis factor receptor 1). MADD can activate TNF-α gene expression and also has the capability to promote the JNK (c-Jun NH2-terminal kinase) pathway. It modulates the recycling of Rab3 small G proteins and plays a major role in Ca2+-dependent neurotransmitter release and exocytosis. This protein also participates in inhibiting the apoptosis of neuronal cells.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST80537

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dual role of DENN/MADD (Rab3GEP) in neurotransmission and neuroprotection.
Miyoshi J and Takai Y
Trends in Molecular Medicine, 10(10), 476-480 (2004)
Engagement of tumor necrosis factor (TNF) receptor 1 leads to ATF-2-and p38 mitogen-activated protein kinase-dependent TNF-α gene expression.
Brinkman BM, et al.
The Journal of Biological Chemistry, 274(43), 30882-30886 (1999)
Regulation of apoptosis and caspase-8 expression in neuroblastoma cells by isoforms of the IG20 gene.
Li L C, et al.
Cancer Research, 68(18), 7352-7361 (2008)
MADD/DENN splice variant of the IG20 gene is necessary and sufficient for cancer cell survival.
Mulherkar N, et al.
Oncogene, 25(47), 6252?6261-6252?6261 (2006)
Wenxiang Bi et al.
Oncology reports, 29(4), 1533-1539 (2013-02-28)
MAPK-activating death domain protein (MADD) binds to the tumor necrosis factor-related apoptosis-inducing ligand (TRAIL) receptor and acts as a key downstream mediator in the TRAIL-induced apoptosis pathway. The aim of this study was to evaluate the expression of MADD in normal human

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service