Skip to Content
Merck
All Photos(8)

Key Documents

HPA031531

Sigma-Aldrich

Anti-CAPZB antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CD85k, Anti-HM18, Anti-ILT3, Anti-LIR-5, Anti-capping protein (actin filament) muscle Z-line, β

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€540.00

€540.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€540.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

€540.00


Please contact Customer Service for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, human, rat

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

CALDLMRRLPPQQIEKNLSDLIDLVPSLCEDLLSSVDQPLKIARDKVVGKDYLLCDYNRDGDSYRSPWSNKYDPP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CAPZB(832)

General description

CAPZB (capping actin protein of muscle Z-line β subunit) gene is mapped to human chromosome 1p36.13. It is widely expressed in pharyngeal arch, and also in lymphoid cells, seminiferous ducts, urothelium and placenta. The gene codes for β-subunit of the barbed-end F-actin-binding protein.

Immunogen

capping protein (actin filament) muscle Z-line, beta recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies®Powered by Atlas Antibodies is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-CAPZB antibody produced in rabbit has been used in immunoblotting and immunofluorescence procedures.

Biochem/physiol Actions

CAPZB (capping actin protein of muscle Z-line βsubunit) is a heterodimeric protein that caps the growing end of F-actin. This actin-capping protein facilitates the joining of actin filaments to the Z-line of the sarcomere in muscles, thereby modulating the cytoskeleton. The protein regulates actin filament dynamics by blocking actin filament assembly and disassembly, and thereby, modulating cell shape and movement in vitro. It is known to promote cell motility by increasing the depolymerization and capping of actin filaments. CAPZB is associated with tumor progression in cases of epithelioid sarcoma.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST76535

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Expression of F-actin-capping protein subunit beta, CAPZB, is associated with cell growth and motility in epithelioid sarcoma.
Mukaihara K
BMC Cancer, 16:206, 1-13 (2016)
Actin capping protein CAPZB regulates cell morphology, differentiation, and neural crest migration in craniofacial morphogenesis?.
Mukherjee K
Human Molecular Genetics, 25(7), 1255-1270 (2016)
Genome-wide association study identifies a novel susceptibility gene for serum TSH levels in Chinese populations.
Zhan M
Human Molecular Genetics, 23(20), 5505-5517 (2014)
Purification and characterization of an alpha 1 beta 2 isoform of CapZ from human erythrocytes: cytosolic location and inability to bind to Mg2+ ghosts suggest that erythrocyte actin filaments are capped by adducin.
Biochemistry, 36(44), 13461-13472 (1997)
Actin capping proteins, CapZ (?-actinin) and tropomodulin in amphioxus striated muscle.
Bao Y
Gene, 510(1), 78-86 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service