Skip to Content
Merck
All Photos(1)

Documents

HPA014048

Sigma-Aldrich

Anti-CYP4F2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CYPIVF2, Anti-Cytochrome P450 4F2, Anti-Cytochrome P450-LTB-omega, Anti-Leukotriene-B(4) 20-monooxygenase, Anti-Leukotriene-B(4) omega-hydroxylase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

RRNWFWGHQGMVNPTEEGMRVLTQLVATYPQGFKVWMGPISPLLSLCHPD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CYP4F2(8529)

General description

The gene CYP4F2 (cytochrome P450 family 4 subfamily F member 2) encodes a protein belonging to the P450 superfamily of enzymes. The gene is mapped to human chromosome 19p13.12. It is expressed in human liver and renal microsomes. It is mostly localized to the S2 and S3 segments of the proximal tubule in kidney.

Immunogen

Cytochrome P450 4F2 recombinant protein epitope signature tag (PrEST)

Application

Anti-CYP4F2 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

The gene CYP4F2 (cytochrome P450 family 4 subfamily F member 2) encodes a Vitamin K1 oxidase that catalyzes the hydroxylation of the tocopherol phytyl side chain in the inactivation pathway of vitamin E. It mediates the ω-hydroxylation of arachidonic acid to 20-HETE (20-Hydroxyeicosatetraenoic acid) in human liver. 20-HETE is involved in the control of kidney vascular and tubular function, vascular tone control in cerebral and coronary circulation, and in turn, blood pressure. Polymorphism in this gene is associated with altered requirement of warfarin dose used to treat thrombotic events. Aberration V433M (rs2108622) in this gene is linked to ischemic stroke in the Northern Chinese Han population.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72369

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael D Caldwell et al.
Blood, 111(8), 4106-4112 (2008-02-06)
Warfarin is an effective, commonly prescribed anticoagulant used to treat and prevent thrombotic events. Because of historically high rates of drug-associated adverse events, warfarin remains underprescribed. Further, interindividual variability in therapeutic dose mandates frequent monitoring until target anticoagulation is achieved.
Matthew G McDonald et al.
Molecular pharmacology, 75(6), 1337-1346 (2009-03-20)
Genetic polymorphisms in VKORC1 and CYP2C9, genes controlling vitamin K(1) (VK1) epoxide reduction and (S)-warfarin metabolism, respectively, are major contributors to interindividual variability in warfarin dose. The V433M polymorphism (rs2108622) in CYP4F2 has also been associated with warfarin dose and
Daniel R Jensen et al.
American journal of medical genetics. Part A, 149A(3), 396-402 (2009-02-14)
We describe a patient with multiple congenital anomalies including deafness, lacrimal duct stenosis, strabismus, bilateral cervical sinuses, congenital cardiac defects, hypoplasia of the corpus callosum, and hypoplasia of the cerebellar vermis. Mutation analysis of EYA1, SIX1, and SIX5, genes that
Shumin Deng et al.
Progress in neuro-psychopharmacology & biological psychiatry, 34(4), 664-668 (2010-03-17)
CYP4F2 is a member of the cytochrome P450 enzymes and is responsible for metabolizing arachidonic acid to 20-hydroxyeicosatetraenoic acid (20-HETE); 20-HETE plays a role in the regulation of vascular tone in the cerebral, coronary, and renal circulation. The present study
J M Lasker et al.
The Journal of biological chemistry, 275(6), 4118-4126 (2000-02-08)
20-hydroxyeicosatetraenoic acid (20-HETE), an omega-hydroxylated arachidonic acid (AA) metabolite, elicits specific effects on kidney vascular and tubular function that, in turn, influence blood pressure control. The human kidney's capacity to convert AA to 20-HETE is unclear, however, as is the

Articles

Phase I biotransformation reactions increase drug compound polarity, mainly occurring in hepatic circulation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service