Skip to Content
Merck
All Photos(1)

Key Documents

C3735

Sigma-Aldrich

Cytochrome P450 human

1A1 Isozyme Microsomes, with P450 Reductase, recombinant, expressed in baculovirus infected insect cells (BTI-TN-5B1-4)

Sign Into View Organizational & Contract Pricing

Select a Size

1 VIAL
€428.00

€428.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 VIAL
€428.00

About This Item

EC Number:
MDL number:
UNSPSC Code:
12161501
NACRES:
NA.47

€428.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

human

Quality Level

recombinant

expressed in baculovirus infected insect cells (BTI-TN-5B1-4)

form

solution

specific activity

≥4 units/pmol enzyme

mol wt

45-60 kDa

packaging

vial of 0.5 nmol

technique(s)

activity assay: suitable

solubility

water: soluble

suitability

suitable for molecular biology

UniProt accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CYP1A1(1543)

Looking for similar products? Visit Product Comparison Guide

General description

Research area: Immuno and CKS

Cytochrome P450 (CYP) enzymes are a family of enzymes that are encoded by the P450 genes. These enzymes are membrane bound hemoproteins. that are mainly found in the liver’s endoplasmic reticulum.

Biochem/physiol Actions

Cytochrome P450 is a heterogeneous family of isozymes whose primary function is to oxidize small molecules, both as a function of intermediary metabolism (e.g., fatty acids) and to detoxify exogenous compounds (drugs or toxins). Some isoforms have narrow substrate specificity, while others are promiscuous. The CYP1A1 isoform catalyzes 7-deethylation of ethoxyresorufin. Cytochrome P450 (CYP) plays an important role in detoxifying xenobiotics, cellular metabolism and homeostasis. One of the main mechanisms of drug-drug interactions is the induction or inhibition of these enzymes. CYP enzymes are transcriptionally activated by a variety of xenobiotics and by endogenous substrates via receptor-dependent pathways. Inhibition of these enzymes is a major factor in metabolism-based drug-drug interactions, and many chemotherapeutic medications can cause drug interactions by either inhibiting or inducing the cytochrome p450 enzyme system.

Physical form

Solution in 100 mM potassium phosphate buffer, pH 7.4.

Preparation Note

Microsomes containing human CYP1A1 and recombinant human NADPH-P450 reductase.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Tomas Erban et al.
Biomedical chromatography : BMC, 26(9), 1062-1065 (2011-11-29)
A 96-well microplate-based HPLC endpoint assay is described for the determination of NADPH-cytochrome P450 reductase (CPR) activity. Novel sampling of NADPH into microplates was optimized. Separation was performed on a Zorbax Eclipse XDB-C₁₈ analytical 4.6 × 150 mm, 5 µm column. To validate the
Palrasu Manikandan et al.
Current drug targets, 19(1), 38-54 (2017-01-27)
The cytochrome P450 (CYP) enzymes are membrane-bound hemoproteins that play a pivotal role in the detoxification of xenobiotics, cellular metabolism and homeostasis. Induction or inhibition of CYP enzymes is a major mechanism that underlies drug-drug interactions. CYP enzymes can be
David Stucki et al.
Archives of biochemistry and biophysics, 687, 108383-108383 (2020-04-27)
Intracellular carbon monoxide (CO) is a gaseous signaling molecule and is generated enzymatically by heme oxygenases upon degradation of heme to billiverdin. Target structures for intracellular produced CO are heme proteins including cytochrome c oxidase of the respiratory chain, cytochrome
D P Bofinger et al.
Toxicological sciences : an official journal of the Society of Toxicology, 62(2), 299-314 (2001-07-14)
Endometriosis is a debilitating disease estimated to affect 10% of reproductive-age women and characterized by the growth of endometrial tissue outside of the uterus. The present study characterizes a human endometrial explant culture model for studying the direct effects of
Moritz Walter et al.
Toxicology in vitro : an international journal published in association with BIBRA, 59, 215-220 (2019-04-21)
Next to its well-studied toxicity, carbon monoxide (CO) is recognized as a signalling molecule in various cellular processes. Thus, CO-releasing molecules (CORMs) are of considerable interest for basic research and drug development. Aim of the present study was to investigate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service