Skip to Content
Merck
All Photos(1)

Key Documents

H8508

Sigma-Aldrich

1,1,1,3,3,3-Hexafluoro-2-propanol

Synonym(s):

HFP, Hexafluoroisopropanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3)2CHOH
CAS Number:
Molecular Weight:
168.04
Beilstein:
1841007
EC Number:
MDL number:
UNSPSC Code:
12352103
Pricing and availability is not currently available.

refractive index

n20/D 1.275 (lit.)

bp

59 °C (lit.)

mp

−4 °C (lit.)

density

1.596 g/mL at 25 °C (lit.)

SMILES string

OC(C(F)(F)F)C(F)(F)F

InChI

1S/C3H2F6O/c4-2(5,6)1(10)3(7,8)9/h1,10H

InChI key

BYEAHWXPCBROCE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service