GE17-0944-01
SOURCE™ 15S
Cytiva 17-0944-01, pack of 50 mL
About This Item
Recommended Products
ligand
sulfonate group
description
Ion Exchanger Type (value)
packaging
pack of 50 mL
manufacturer/tradename
Cytiva 17-0944-01
storage condition
dry at room temperature
matrix
polystyrene/divinylbenzene
avg. part. size
15 μm
particle size
15-15 μm
cleaning in place
1-14
working range
2-13
suitability
suitable for bioprocess medium (BioProcess medium. Process-scale quantities are available.)
suitable for bioprocess medium
Related Categories
General description
Application
With its very high resolution, SOURCE™ 15S is suitable for polishing steps in both research and industrial applications.
Through the combination of high quality assurance standards and a patented manufacturing process, particle structure is consistent both bead-to-bead and batch-to batch.
As member of the BioProcess media range, SOURCE™ 15S meets industrial demands with security of supply and comprehensive technical and regulatory support.
Features and Benefits
- Mono-sized 15 um rigid beads gives Low back pressure and high flow rates allowing for high productivity
- Strong cation exchanger designed for high resolution polishing purification of Proteins, peptides and oligonucleotides
- High chemical stability allowing for wide range of working conditions with good resistance to cleaning conditions at high pH
Storage and Stability
Analysis Note
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Storage Class Code
3 - Flammable liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
This page shows Hydrophobic Interaction Chromatography (HIC) in a purification strategy.
Column, media, and sample preparation for Hydrophobic Interaction Chromatography (HIC) detailed with Cytiva media.
Protocols
In this section the practical aspect of Reverse Phased Chromatography ( RPC) is discussed including media and column selection and eluent selection and preparation.
Selectivity of HIC medium from Cytiva is based on properties such as hydrophobicity, porosity and particle size.
This page discusses how to do a purification with SOURCE™ media from Cytiva.
This page discusses various aspects of sample preparation for chromatographic purification.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service