Skip to Content
Merck
All Photos(2)

Documents

GE17-0404-01

HiTrap® Protein G High Performance

Cytiva 17-0404-01, pack of 5 × 1 mL

Synonym(s):

Protein purification column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151816
NACRES:
NA.56

ligand

recombinant protein G lacking albumin-binding region

packaging

pack of 5 × 1 mL

manufacturer/tradename

Cytiva 17-0404-01

storage condition

(20% Ehtanol)
(20% Ethanol)

parameter

<4 mL/min flow rate
42 psi

bed size

7 mm × 25 mm

bed volume

1 mL

column I.D.

7 mm

size

avg. part. size

34 μm

cleaning

2-9

working range

3-9

capacity

>1 mg binding capacity (lgG/column)
25 mg binding capacity(human IgG/column)

storage temp.

2-8°C

Related Categories

General description

HiTrap® Protein G HP columns are prepacked with Protein G Sepharose High Performance for fast, easy separations of IgG from different species and subclasses.
The pressure over the packed bed varies depending on a range of parameters such as the characteristics of the chromatography medium and the column tubing used.
pH below 3 is sometimes required to elute strongly bound IgG species. However, protein ligands may hydrolyze at very low pH.
pH below 3 is sometimes required to elute strongly bound IgG species. However, protein ligands may hydrolyze at very low pH.

Features and Benefits

  • High binding capacity of 25 mg IgG/mL medium and fast binding kinetics.
  • 34 μm bead size for high performance, high-resolution purifications.
  • Binds broad range of different species and subclasses of IgG.
  • Simple and proven method giving reproducible results.
  • Designed for use with syringe, peristalic pump, or chromatography system.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Other Notes

The pressure over the packed bed varies depending on a range of parameters such as the characteristics of the chromatography medium and the column tubing used.

Legal Information

HiTrap is a registered trademark of Cytiva
Sepharose is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lin-Ping Wu et al.
Nature communications, 12(1), 4858-4858 (2021-08-13)
Complement is an enzymatic humoral pattern-recognition defence system of the body. Non-specific deposition of blood biomolecules on nanomedicines triggers complement activation through the alternative pathway, but complement-triggering mechanisms of nanomaterials with dimensions comparable to or smaller than many globular blood

Articles

Designing purification methods for antibodies ensures quality and quantity adequacy for research and industrial purposes.

Purify monoclonal or polyclonal IgG from serum, cell culture supernatant or ascitic fluid using the HiTrap Protein G HP from Cytiva, an affinity-exclusion chromatography product containing Sepharose-immobilized Protein G.

This page shows how to purify IgM antibodies by affinity chromatography using HiTrap™ IgM Purification HP.

This page shows how to separate IgG antibodies by affinity chromatography using HiTrap® Protein G HP columns from Cytiva.

See All

Protocols

This page shows how to perform a two-step purification of mouse monoclonal IgG1 using HiTrap® Protein G HP from Cytiva for the capture step.

General instructions for affinity purification using HiTrap columns from Cytiva.

This page shows how to prepare samples for purification with affinity chromatography.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service