Skip to Content
Merck
All Photos(2)

Key Documents

GE29-0485-76

HiTrap® Protein A High Performance

Cytiva 29-0485-76, pack of 1 mL

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
€197.00

€197.00


Please contact Customer Service for Availability


Select a Size

Change View
1 PKG
€197.00

About This Item

UNSPSC Code:
23151816
NACRES:
NA.56

€197.00


Please contact Customer Service for Availability

packaging

pack of 1 mL

manufacturer/tradename

Cytiva 29-0485-76

availability

not available in North America

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

HiTrap Protein A HP is a prepacked ready to use, column for preparative purification of monoclonal and polyclonal antibodies. The special design of the column, together with the medium provide fast, simple and easy separations in a convenient format.

Application

HiTrap Protein A HP is a prepacked Protein A Sepharose High Performance column for preparative purification of monoclonal and polyclonal antibodies from ascites, serum, and cell culture supernatants.

Features and Benefits

  • High binding capacitiy of 20 mg IgG/mL medium and fast kinetics.
  • 34 μm bead size for high-performance, high-resolution purifications.
  • Designed for use with syringe, pump, or chromatography system such as ÄKTA design or FPLC System.
  • Stable over pH 3 to 9 for use, and 2 to 10 for cleaning.
  • Robust, easy-to-use method.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Legal Information

HiTrap is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This page shows how to purify IgM antibodies by affinity chromatography using HiTrap™ IgM Purification HP.

This page shows how to separate IgG antibodies by affinity chromatography using HiTrap® Protein G HP columns from Cytiva.

This page shows how to solve practical problems that may occur when running an affinity chromatography column.

Protocols

General instructions for affinity purification using HiTrap columns from Cytiva.

Protein A, coupled to Sepharose, binds IgG via its Fc regions, facilitating affinity chromatography purification.

This page shows how to convert between linear flow and volumetric flow rates in affinity chromatography.

How to perform buffer exchange and desalting with Sephadex G-25, HiTrap Desalting columns, or ÄKTAprime plus.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service