Skip to Content
Merck
All Photos(1)

Key Documents

GE17-0160-01

Sepharose Cl-6B

Cytiva 17-0160-01, pack of 1 L

Synonym(s):

Ion-exchange resin, Sepharose gel

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
€1,090.00

€1,090.00


Please contact Customer Service for Availability


Select a Size

Change View
1 PKG
€1,090.00

About This Item

UNSPSC Code:
41106500
NACRES:
NA.56

€1,090.00


Please contact Customer Service for Availability

feature

autoclavable Autoclavable, 20 min at 120°C in pH 7

packaging

pack of 1 L

manufacturer/tradename

Cytiva 17-0160-01

matrix

6% agarose

particle size

45-165 μm

cleaning in place

2-14

working range

3-13

Related Categories

General description

Sepharose CL-6B is a well-proven cross-linked agarose gel filtration base matrix and is frequently used for coupling affinity ligands to the matrix. The matrix is not pre-activated and the user performs all steps in coupling.

Sepharose is a bead-formed agarose-based gel filtration matrix. Sepharose is available with 3 different agarose contents; 2, 4, and 6%, designated Sepharose 2B, Sepharose 4B and Sepharose 6B respectively, Both Sepharose and Sepharose CL have broad fractionation ranges which makes them suitable for characterizing or cleaning-up samples containing components of diverse molecular weight.

Sepharose CL gels are cross-linked derivatives of Sepharose 2B, Sepharose 4B and Sepharose 6B. The cross-linked form of Sepharose is chemically and physically more resistant than Sepharose itself, offering the same selectivity with better flow characteristics. Cross-linked Sepharose gels are resistant to organic solvents and are thus the choice for separations in organic solvents.

Features and Benefits

  • 6% cross-linked agarose gel filtration media.
  • Proven base matrix for coupling affinity ligands
  • Higher degree of crosslinking improve matrix stability

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Sepharose is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service