Skip to Content
Merck
All Photos(1)

Documents

GE17-0150-01

Sepharose CL-4B

Cytiva 17-0150-01, pack of 1 L

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41106500
NACRES:
NA.56

feature

autoclavable Autoclavable, 20 min at 120°C in pH 7

packaging

pack of 1 L

manufacturer/tradename

Cytiva 17-0150-01

matrix

4% agarose

particle size

45-165 μm

cleaning in place

2-14

working range

3-13

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Sepharose CL-4B is a well-proven cross-linked agarose gel filtration base matrix and is frequently used for coupling affinity ligands to the matrix. The matrix is not pre-activated and the user performs all steps in coupling.

Sepharose is a bead-formed agarose-based gel filtration matrix. Sepharose is available with 3 different agarose contents; 2, 4, and 6%, designated Sepharose 2B, Sepharose 4B and Sepharose 6B respectively, Both Sepharose and Sepharose CL have broad fractionation ranges which makes them suitable for characterizing or cleaning-up samples containing components of diverse molecular weight.

Sepharose CL gels are cross-linked derivatives of Sepharose 2B, Sepharose 4B and Sepharose 6B. The cross-linked form of Sepharose is chemically and physically more resistant than Sepharose itself, offering the same selectivity with better flow characteristics. Cross-linked Sepharose gels are resistant to organic solvents and are thus the choice for separations in organic solvents.

Features and Benefits

  • 4% cross-linked agarose gel filtration media.
  • Proven base matrix for coupling affinity ligands
  • Higher degree of crosslinking improve matrix stability

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Sepharose is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Protocols

Selectivity of HIC medium from Cytiva is based on properties such as hydrophobicity, porosity and particle size.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service