Skip to Content
Merck
All Photos(1)

Key Documents

G8291

Sigma-Aldrich

Glu-Val-Asn-[(2R,4S,5S)-5-amino-4-hydroxy-2,7-dimethyloctanoyl]-Ala-Glu-Phe

>93% (HPLC), solid

Synonym(s):

OM99-2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H64N8O14
CAS Number:
Molecular Weight:
892.99
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

>93% (HPLC)

form

solid

solubility

0.1% trifluoroacetic acid: 1 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

CC(C)C[C@H](NC(=O)[C@H](CC(N)=O)NC(=O)[C@@H](NC(=O)[C@@H](N)CCC(O)=O)C(C)C)[C@@H](O)C[C@@H](C)C(=O)N[C@@H](C)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](Cc1ccccc1)C(O)=O

InChI

1S/C41H64N8O14/c1-20(2)16-27(46-39(60)28(19-31(43)51)47-40(61)34(21(3)4)49-37(58)25(42)12-14-32(52)53)30(50)17-22(5)35(56)44-23(6)36(57)45-26(13-15-33(54)55)38(59)48-29(41(62)63)18-24-10-8-7-9-11-24/h7-11,20-23,25-30,34,50H,12-19,42H2,1-6H3,(H2,43,51)(H,44,56)(H,45,57)(H,46,60)(H,47,61)(H,48,59)(H,49,58)(H,52,53)(H,54,55)(H,62,63)/t22-,23+,25+,26+,27+,28+,29+,30+,34+/m1/s1

InChI key

NIOCOJFPGCXNKL-ZKZXETMPSA-N

Amino Acid Sequence

Glu-Val-Asn-[(2R,4S,5S)-5-amino-4-hydroxy-2,7-dimethyloctanoyl]-Ala-Glu-Phe

Biochem/physiol Actions

Inhibitor of brain β-secretase

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Design of potent inhibitors for human brain memapsin (β-secretase).
Ghosh, A.K.
Journal of the American Chemical Society, 122, 3522-3522 (2000)
L Hong et al.
Science (New York, N.Y.), 290(5489), 150-153 (2000-10-06)
Memapsin 2 (beta-secretase) is a membrane-associated aspartic protease involved in the production of beta-amyloid peptide in Alzheimer's disease and is a major target for drug design. We determined the crystal structure of the protease domain of human memapsin 2 complexed
Mohammad Mahdavi et al.
Chemistry & biodiversity, 16(4), e1800488-e1800488 (2019-02-06)
Alzheimer's disease (AD) is a well-known neurodegenerative disorder affecting millions of old people worldwide and the corresponding epidemiological data emphasize the importance of the disease. As AD is a multifactorial illness, various single target directed drugs that have reached clinical
Donatella Panatto et al.
Human vaccines & immunotherapeutics, 11(1), 270-276 (2014-12-09)
Infections caused by Chlamydia trachomatis (Ct) and human papillomavirus (HPV) are the two main sexually transmitted infections; however, epidemiological data on Ct prevalence and Ct/HPV co-infection in Italy are scant. This study aimed at estimating the prevalence of Ct infection
Ramin Miri et al.
Archives of pharmacal research, 38(4), 456-469 (2014-04-29)
β-site amyloid precursor protein cleaving enzyme (BACE-1) is a validated target for Alzheimer therapy due to its distinctive role in pathogenesis of AD. In the present contribution, a series of new 3,5-bis-N-(aryl/heteroaryl) carbamoyl-4-aryl-1,4-dihydropyridine structures were synthesized as BACE-1 inhibitors (6a-6n).

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service