Merck
  • Inhibitors of Alzheimer's BACE-1 with 3,5-bis-N-(aryl/heteroaryl) carbamoyl-4-aryl-1,4-dihydropyridine structure.
Inhibitors of Alzheimer's BACE-1 with 3,5-bis-N-(aryl/heteroaryl) carbamoyl-4-aryl-1,4-dihydropyridine structure.
Archives of pharmacal research (2014-04-29)
Ramin Miri, Omidreza Firuzi, Nima Razzaghi-Asl, Katayoun Javidnia, Najmeh Edraki
ABSTRACT
β-site amyloid precursor protein cleaving enzyme (BACE-1) is a validated target for Alzheimer therapy due to its distinctive role in pathogenesis of AD. In the present contribution, a series of new 3,5-bis-N-(aryl/heteroaryl) carbamoyl-4-aryl-1,4-dihydropyridine structures were synthesized as BACE-1 inhibitors (6a-6n). In vitro BACE-1 inhibitory activities were determined by enzymatic fluorescence resonance energy transfer assay. Synthesized dihydropyridine (DHP) analogues exhibited weak to good inhibitory activities while 6i, 6n and 6a were found to be the most potent molecules with 83.76, 79.45 and 72.47 % BACE-1 inhibition at 10 μM, respectively. Structure binding/activity relationship elucidations revealed that superior BACE-1 inhibitory activities were observed for DHP derivatives bearing fused/non-fused thiazole groups and particularly 3,5-bis-N-(6-ethoxy-2-benzothiazolyl) moiety. Binding maps showed that enhanced activity may be attributed to the additional H-bond and hydrophobic interactions with S2-S3 subpockets of BACE-1.
Sigma-Aldrich
Potassium bromide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Potassium bromide, FT-IR grade, ≥99% trace metals basis
Sigma-Aldrich
Potassium bromide, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium bromide, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.95% trace metals basis
Sigma-Aldrich
Potassium bromide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Potassium bromide, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium bromide, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%