Skip to Content
Merck
All Photos(1)

Key Documents

EP5702742007

Eppendorf® 20 Rubber Mats

for use with Adapter 5702 742.007, pack of 20 ea

Synonym(s):

Rubber Mat for Eppendorf® A-4-38 Rotor

Sign Into View Organizational & Contract Pricing

Select a Size

20 EA
€37.80

€37.80


Please contact Customer Service for Availability


Select a Size

Change View
20 EA
€37.80

About This Item

UNSPSC Code:
41103911
NACRES:
NB.82

€37.80


Please contact Customer Service for Availability

feature

Holds 10 mL tubes

packaging

pack of 20 ea

manufacturer/tradename

Eppendorf® 5702742007

technique(s)

centrifugation: suitable

compatibility

for use with Adapter 5702 742.007

Looking for similar products? Visit Product Comparison Guide

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service