EP0030119479
Eppendorf Tube®
capacity 5.0 mL, Biopur®, pack of 50 ea (individually wrapped)
Synonym(s):
eppendorf tubes, microcentrifuge tubes, microfuge tubes
Select a Size
€58.30
Select a Size
About This Item
€58.30
Recommended Products
material
cap (snap cap)
colorless cap
colorless tube
conical bottom
polypropylene cap
polypropylene tube
transparent polypropylene
sterility
sterile
feature
graduations
neck thread Push fit
Biopur®
conical shaped bottom with hinged lid
packaging
pack of 50 ea (individually wrapped)
manufacturer/tradename
Eppendorf® 0030119479
parameter
25,000 × g
capacity
5.0 mL
Looking for similar products? Visit Product Comparison Guide
General description
- Maximum safety and stability for centrifugation up to 25,000 × g
- Conical bottoms
- Easier processing of samples up to 5.0 mL
- Higher yields in DNA isolation, prep of mastermixes and buffers, and safe cell and tissue lysis
- Transparent polypropylene, free of plasticizers, biocides or mold release agents
- Hinged lid for minimized sample evaporation during storage and incubation in a wide range of temperatures from -86°C to 80°C
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service