EP0030089553
Eppendorf® Combitips Advanced
volume 2.5 mL, green, sterile, pack of 100 ea (individually wrapped)
Synonym(s):
pipetting tips
All Photos(1)
About This Item
UNSPSC Code:
41121600
volume:
2.5 mL
sterility:
sterile
feature:
graduation: no
packaging:
pack of 100 ea (individually wrapped)
manufacturer/tradename:
Eppendorf® 0030089553
Recommended Products
material
green
polypropylene
sterility
sterile
feature
graduation: no
packaging
pack of 100 ea (individually wrapped)
manufacturer/tradename
Eppendorf® 0030089553
volume
2.5 mL
Related Categories
General description
Eppendorf Combitips are our tip of choice when working with Multipette multiple dispensing pipettes. Combitips advanced dispenser tips offer you many advantages for working in a modern laboratory thanks to their revolutionary design. With an integrated piston, they work according to the positive displacement principle and ensure that the correct volume of liquid is dispensed regardless of the physical properties of the liquid you are working with (e.g. viscosity, volatility, density, temperature). This allows you to dispense with high precision regardless of the flow properties of the liquid and, thanks to the hermetically sealed piston that prevents contamination by aerosols, you can work safely with radioactive or toxic materials. Eppendorf pioneered the Multipette/Combitips multiple dispensing system in 1978 and has improved it several times since then. This was the very first pipetting system that allowed the user to fill the tip once and dispense multiple times. The concept continues to be a driver for efficient liquid handling and is the tool of choice in many laboratories working with plates or series of reaction vessels. In 2023, the Combitips advanced pipette tips were certified with ACT Environmental Impact Factor labels by the non-profit organization My Green Lab . This makes them the ideal choice for laboratories looking for more sustainable products.
Features and Benefits
- Direct displacement principle (similar to a syringe)
- High-precision dispensing regardless of the physical properties of the liquid (e.g. viscosity, volatility, density or temperature)
- Prevents aerosol contamination through the hermetically sealed piston and thus ensures safe dispensing
- Protection against radioactive and toxic substances
- Fast dispensing of long series through precise, repeated dosing of identical volumes (in combination with Multipette hand dispensers)
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)