EP0030076265
Eppendorf® Empty Box 2.0 for epT.I.P.S.® Reloads
for tips 1250 µL – 5 mL
Select a Size
€41.10
Select a Size
About This Item
€41.10
Recommended Products
feature
for tips 1250 µL – 5 mL
packaging
pkg of 1 ea
manufacturer/tradename
Eppendorf® 0030076265
volume
1250—5000 μL
General description
Application
Features and Benefits
- epT.I.P.S.® Box 2.0 – empty reusable boxes for tip volumes of 10 µL - 5 mL
- epT.I.P.S.® Box 2.0 – all three sizes are availabe in Eppendorf Quality
- Autoclavable at 121 °C (20 min.) for subsequent use
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)