EP0030076010
Eppendorf® epT.I.P.S.® G Reload 384
PCR clean, tip capacity × L 0.5-100 μL × 53 mm, light yellow tip, case of 3,840 ea (10 trays x 384 tips)
Synonym(s):
pipetting tips
Select a Size
€415.00
Select a Size
About This Item
€415.00
Recommended Products
sterility
non-sterile
feature
PCR clean
packaging
case of 3,840 ea (10 trays x 384 tips)
manufacturer/tradename
Eppendorf® 0030076010
tip capacity × L
0.5-100 μL × 53 mm
volume
100 μL
color
light yellow tip
storage temp.
room temp
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Designed for a cone distance of 4.5 mm
- epT.I.P.S. 384 pipette tips build a system together with the Research plus and Xplorer plus 16 and 24 channel pipettes for utmost safety and reliability
- Innovative SOFTattach technology allows for reliable tip fit and tightness
- Fill a whole 384 well plate without loosening pipette tips
- Perfect tip alignment by extraordinary coaxiality for safe plate maneuvering and liquid transfer
- The tip shape of the epT.I.P.S. 384 is adjusted to perfectly fit the wells of 384 well plates
- Tip attachment forces reduced by 40% per cone compared to 8 and 12 channel pipettes
- Tip ejection forces reduced by SOFTattach for ergonomic working conditions
- Available as reload, and set
- Available in purity grades: Eppendorf Quality and PCR clean
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service