Skip to Content
Merck
All Photos(1)

Key Documents

EHU100421

Sigma-Aldrich

MISSION® esiRNA

targeting human FGFR1 (1)

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€192.00
50 μG
€341.00

€192.00


Please contact Customer Service for Availability


Select a Size

Change View
20 μG
€192.00
50 μG
€341.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

€192.00


Please contact Customer Service for Availability

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCCTCTGACAAGGGCAACTACACCTGCATTGTGGAGAATGAGTACGGCAGCATCAACCACACATACCAGCTGGATGTCGTGGAGCGGTCCCCTCACCGGCCCATCCTGCAAGCAGGGTTGCCCGCCAACAAAACAGTGGCCCTGGGTAGCAACGTGGAGTTCATGTGTAAGGTGTACAGTGACCCGCAGCCGCACATCCAGTGGCTAAAGCACATCGAGGTGAATGGGAGCAAGATTGGCCCAGACAACCTGCCTTATGTCCAGATCTTGAAGACTGCTGGAGTTAATACCACCGACAAAGAGATGGA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fei Ma et al.
Journal of experimental & clinical cancer research : CR, 36(1), 158-158 (2017-11-15)
MicroRNAs function as key regulators in various human cancers, including breast cancer (BC). MiR-361-5p has been proved to be a tumor suppressor in colorectal cancer and gastric cancer in our previous study. In this study, we aim to find out
Jiexia Zhang et al.
International journal of oncology, 54(6), 2211-2221 (2019-04-04)
Emerging reports have revealed that several microRNAs (miRNAs) are abnormally expressed in non‑small cell lung cancer (NSCLC). miRNAs have been identified as oncogenes or tumor suppressors, and regulate various biological processes including oncogenesis and development. miR‑802 is dysregulated in multiple
Sheyda Azimi et al.
Microbial pathogenesis, 149, 104534-104534 (2020-10-13)
Neisseria meningitidis (the meningococcus) remains an important cause of human disease, including meningitis and sepsis. Adaptation to the host environment includes many interactions with specific cell surface receptors, resulting in intracellular signalling and cytoskeletal rearrangements that contribute to pathogenesis. Here
Liyan Wang et al.
FEBS letters, 590(23), 4252-4262 (2016-10-23)
MiR-296 was previously reported to be underexpressed in hepatocellular carcinoma (HCC). However, the clinical value of miR-296 and its function in HCC remain poorly understood. In this study, we found that miR-296 levels are decreased in HCC specimens and cells
Tetsuya Kawane et al.
Scientific reports, 8(1), 13551-13551 (2018-09-12)
Runx2 and Sp7 are essential transcription factors for osteoblast differentiation. However, the molecular mechanisms responsible for the proliferation of osteoblast progenitors remain unclear. The early onset of Runx2 expression caused limb defects through the Fgfr1-3 regulation by Runx2. To investigate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service