Skip to Content
Merck
All Photos(1)

Key Documents

EHU091111

Sigma-Aldrich

MISSION® esiRNA

targeting human UBE2K

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€192.00
50 μG
€341.00

€192.00


Please contact Customer Service for Availability


Select a Size

Change View
20 μG
€192.00
50 μG
€341.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

€192.00


Please contact Customer Service for Availability

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TCCGCACGGTATTATTGTCATTGCAAGCACTATTGGCAGCTGCAGAGCCAGATGATCCACAGGATGCTGTAGTAGCAAATCAGTACAAACAAAATCCCGAAATGTTCAAACAGACAGCTCGACTTTGGGCACATGTGTATGCTGGAGCACCAGTTTCTAGTCCAGAATACACCAAAAAAATAGAAAACCTATGTGCTATGGGCTTTGATAGGAATGCAGTAATAGTGGCCTTGTCTTCAAAATCATGGGATGTAGAGACTGCAACAGAATTGCTTCTGAGTAACTGAGGCATAGAGAGCTGCTGATATAGTCAAGCTTGCCTCTTCTTGAGGAGCACCAACATCTGTTATTTTTAGGATTCTGCATAGATTTCTTTTAAACTGGCATTCTTGCCTAATGATGTTATCTAGGCACCATTGGAGACTG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eun Il Jeong et al.
Cell death & disease, 7(12), e2573-e2573 (2016-12-30)
Cerebral ischemia/reperfusion (I/R) causes brain damage accompanied by ubiquitin accumulation and impairment of proteasome activity. In this study, we report that E2-25K, an E2-conjugating enzyme, is SUMOylated during oxidative stress and regulates cerebral I/R-induced damage. Knockdown of E2-25K expression protects
Wen-Bin Gu et al.
Developmental and comparative immunology, 101, 103452-103452 (2019-07-19)
NFIL3 is a transcriptional activator of the IL-3 promoter in T cells. In vertebrates, it has been characterized as an essential regulator of several cellular processes such as immunity response, apoptosis and NK cells maturation. However, the identification and functional
Lisa Schmölz et al.
Biochimica et biophysica acta, 1863(8), 919-927 (2018-05-08)
The long-chain metabolites of vitamin E (LCM) emerge as a new class of regulatory metabolites and have been considered as the active compounds formed during vitamin E metabolism. The bioactivity of the LCM is comparable to the already established role

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service