Skip to Content
Merck
All Photos(1)

Key Documents

EHU002711

Sigma-Aldrich

MISSION® esiRNA

targeting human BIRC2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GTGGCCTGATGCTGGATAACTGGAAACTAGGAGACAGTCCTATTCAAAAGCATAAACAGCTATATCCTAGCTGTAGCTTTATTCAGAATCTGGTTTCAGCTAGTCTGGGATCCACCTCTAAGAATACGTCTCCAATGAGAAACAGTTTTGCACATTCATTATCTCCCACCTTGGAACATAGTAGCTTGTTCAGTGGTTCTTACTCCAGCCTTTCTCCAAACCCTCTTAATTCTAGAGCAGTTGAAGACATCTCTTCATCGAGGACTAACCCCTACAGTTATGCAATGAGTACTGAAGAAGCCAGATTTCTTACCTACCATATGTGGCCATTAACTTTTTTGTCACCATCAGAATTGGCAAGAGCTGGTTTTTATTATATAGGACCTGGAGATAGGGTAGCCTGCTTTGCCTGTGGTGGGAAGCTCAGTAACTGGGAACCAAAGGATGATGCTATGTCAGAACACCGGAGGCATTTTCCCAACTGTCCATTTTTGGAAAATTCTCTAGAAACTCTGAGGTTTAGCATTTCAAATCTGAGCATGCAGACACATGCAGCTCGAAT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xin Wang et al.
International journal of developmental neuroscience : the official journal of the International Society for Developmental Neuroscience, 53, 10-17 (2016-05-26)
Bone morphogenic protein-7 (BMP7) is a multifunctional cytokine with demonstrated neurogenic potential. Oligodendrocytes (OLs) death after spinal cord injury (SCI) contributes to demyelination of spared axons, even leading to a permanent neurological deficit. Therefore, therapeutic approaches to prevent OLs death
M Lappas
Placenta, 35(10), 831-838 (2014-08-13)
Independent of their role in apoptosis, cellular inhibitors of apoptosis (cIAP) 1 and 2, have emerged as regulators of inflammation. Obesity in pregnancy is characterised by maternal and placental inflammation. Thus, the aim of this study was to determine the
Franziska Mueller et al.
Science advances, 7(3) (2021-02-02)
SMAC/DIABLO and HTRA2 are mitochondrial proteins whose amino-terminal sequences, known as inhibitor of apoptosis binding motifs (IBMs), bind and activate ubiquitin ligases known as inhibitor of apoptosis proteins (IAPs), unleashing a cell's apoptotic potential. IBMs comprise a four-residue, loose consensus
Hong Jin et al.
Oncology research, 22(3), 167-176 (2015-07-15)
Emerging evidence suggests a potential role of cellular inhibitor of apoptosis protein 1 (cIAP1) in the development of human ovarian cancer. However, its function in the progression of ovarian cancer has not been clearly determined. Our study aimed to investigate
E-W Lee et al.
Cell death and differentiation, 22(9), 1463-1476 (2015-01-24)
Given their crucial role in apoptosis suppression, inhibitor of apoptosis proteins (IAPs) have recently become attractive targets for cancer therapy. Here, we report that cellular IAP2 (cIAP2) is specifically stabilized in several cancer cell lines, leading to resistance to Smac

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service