Merck
  • Overlap of NatA and IAP substrates implicates N-terminal acetylation in protein stabilization.
Overlap of NatA and IAP substrates implicates N-terminal acetylation in protein stabilization.
Science advances (2021-02-02)
Franziska Mueller, Alexandra Friese, Claudio Pathe, Richard Cardoso da Silva, Kenny Bravo Rodriguez, Andrea Musacchio, Tanja Bange
ABSTRACT
SMAC/DIABLO and HTRA2 are mitochondrial proteins whose amino-terminal sequences, known as inhibitor of apoptosis binding motifs (IBMs), bind and activate ubiquitin ligases known as inhibitor of apoptosis proteins (IAPs), unleashing a cell's apoptotic potential. IBMs comprise a four-residue, loose consensus sequence, and binding to IAPs requires an unmodified amino terminus. Closely related, IBM-like N termini are present in approximately 5% of human proteins. We show that suppression of the N-alpha-acetyltransferase NatA turns these cryptic IBM-like sequences into very efficient IAP binders in cell lysates and in vitro and ultimately triggers cellular apoptosis. Thus, amino-terminal acetylation of IBM-like motifs in NatA substrates shields them from IAPs. This previously unrecognized relationship suggests that amino-terminal acetylation is generally protective against protein degradation in human cells. It also identifies IAPs as agents of a general quality control mechanism targeting unacetylated rogues in metazoans.
Sigma-Aldrich
MISSION® esiRNA, targeting human XIAP
Sigma-Aldrich
MISSION® esiRNA, targeting human BIRC2
Sigma-Aldrich
L-Arginine monohydrochloride, not synthetic, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human NAA20
BRAND® 96-well microplate, U-bottom, round bottom, non-sterile
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%