Skip to Content
Merck
All Photos(1)

Key Documents

E003876

Sigma-Aldrich

BICINE

Synonym(s):

N,N-Bis(2-hydroxyethyl)glycine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13NO4
CAS Number:
Molecular Weight:
163.17
Beilstein:
1769362
EC Number:
MDL number:
UNSPSC Code:
12161700

Pricing and availability is not currently available.

useful pH range

7.6-9.0

pKa (25 °C)

8.3

SMILES string

OCCN(CCO)CC(O)=O

InChI

1S/C6H13NO4/c8-3-1-7(2-4-9)5-6(10)11/h8-9H,1-5H2,(H,10,11)

InChI key

FSVCELGFZIQNCK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Recommended buffer for low temperature biochemical work. Used in the preparation of stable substrate solution for serum guanase determination. Spacer in plasma protein fractionation with Sephadex® by isotachophoresis.

Legal Information

Sephadex is a registered trademark of Cytiva

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Irradiation inactivation analysis of acetylcholinesterase and the effect of buffer salts.
D Parkinson et al.
Radiation research, 90(2), 252-259 (1982-05-01)
Taufika Islam Williams et al.
Electrophoresis, 27(14), 2984-2995 (2006-05-24)
A novel, Bicine-based SDS-PAGE buffer system was developed for the analysis of membrane proteins. The method involves molecular weight-based separations of fully denatured and solubilized proteins in two dimensions. This doubled SDS-PAGE (dSDS-PAGE) approach produced a diagonal arrangement of protein
E Gopinath et al.
Proceedings of the National Academy of Sciences of the United States of America, 86(9), 3041-3044 (1989-05-01)
The nature and rate of reduction of Hg2+ to Hg0 by 1,5-dihydro-3,(3-sulfopropyl)lumiflavin (FIH2) in buffered aqueous solutions (pH 4.7) is dependent on the ligation of Hg2+. In the presence of N,N-bis(2-hydroxyethyl)glycine or when ligated to ethylenediaminetetraacetic acid, the reduction is
The myosin dimer: an intermediate in the self-assembly of the thick filament of vertebrate skeletal muscle.
J S Davis et al.
FEBS letters, 140(2), 293-297 (1982-04-19)
B M Altura et al.
British journal of pharmacology, 69(2), 207-214 (1980-06-01)
1 In vitro studies were undertaken on rat aortic strips and portal vein segments in order to determine whether or not several commonly used artificial buffers, i.e., tris(hydroxymethyl) aminomethane (Tris), N-2-hydroxyethylpiperazine-N'-2-ethanesulphonic acid (HEPES), morpholine propanesulphonic acid (MOPS), N,N bis(2-hydroxyethyl) glycine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service