Skip to Content
Merck
All Photos(1)

Documents

CLS432014

Corning® XT Starter

complete, low profile

Synonym(s):

Starter Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103800
NACRES:
NB.12

feature

complete, low profile

packaging

pkg of 1

manufacturer/tradename

Corning 432014

L × W × H

16.2 cm × 12.1 cm × 6.1 cm

color

purple

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

An open-platform ice-free cooler that accommodates most CoolRack and CoolSink modules. Low profile and small footprint make it ideal for use in the hood, keeping samples cold (0.5° to 4.0°C) up to four hours. 1°C to 8°C temperature indicator provides visual assurance of temperature performance. To extend the cooling duration, keep an additional XT Cooling Core in the freezer and rotate the Cores as needed.

Legal Information

Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service