Skip to Content
Merck
All Photos(1)

Key Documents

CLS3642

Corning® 96 Well Half-Area Microplate

flat bottom, white polystyrene, bag of 25 inner package, non-sterile, lid: no

Synonym(s):

cell culture plate, multi well plates, multiple well plates

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
€1,260.00

€1,260.00


Please contact Customer Service for Availability


Select a Size

Change View
100 EA
€1,260.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.24
material:
flat bottom
round wells
white polystyrene
white polystyrene plate
size:
96 wells

€1,260.00


Please contact Customer Service for Availability

material

flat bottom
round wells
white polystyrene
white polystyrene plate

sterility

non-sterile

feature

lid: no
skirt

packaging

bag of 25 inner package

manufacturer/tradename

Corning 3642

maximum volume

190 μL

size

96 wells

well D

10.7 mm

working volume

125 μL

color

white

suitability

suitable for (cell cuture)

binding type

NBS (non-binding surface)
low binding surface

Looking for similar products? Visit Product Comparison Guide

General description

Unique half area plates can help save on rare and expensive reagents by reducing the amount of reagent needed per well, while still being readable by most standard plate readers.

Features and Benefits

  • Corning® 96 Well Half-Area Microplate flat bottom white polystyrene enhances luminescent signals and has low background fluorescence.
  • Display minimal well-to-well crosstalk.
  • Recommended working volume of up to 20 to 80 μL.
  • Ideal for reducing protein and nucleic acid binding at low concentrations.
  • Without lids, lids are supplied separately.
  • Conforms to standard microplate footprint and dimensions.

Legal Information

Corning is a registered trademark of Corning, Inc.
NBS is a trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service