Skip to Content
Merck
All Photos(1)

Key Documents

AB7144F

Sigma-Aldrich

Anti-Fibrinogen Antibody, FITC conjugated

Chemicon®, from sheep

Sign Into View Organizational & Contract Pricing

Select a Size

2 ML
€351.00

€351.00


Estimated to ship on07 April 2026Details


Request a Bulk Order

Select a Size

Change View
2 ML
€351.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€351.00


Estimated to ship on07 April 2026Details


Request a Bulk Order

biological source

sheep

Quality Level

conjugate

FITC conjugate

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunodiffusion: suitable
immunofluorescence: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FIBCD1(84929)

Specificity

Prior to conjugation, the antibody was specific for fibrinogen when tested against human plasma by Coomassie Blue R stained double immunodiffusion and immunoelectrophoresis. Undesired immune reactivity has been removed by solid phase immunoadsorption.

LABELLING RATIO

Fluorescein:antibody molar ratio: 2.8 - 3.3

Absorbance ratio (A493/A276): 0.78 + 0.05

Immunogen

Purified fibrinogen derived from pooled human plasma.

Application

Detect Fibrinogend using this Anti-Fibrinogen Antibody, FITC conjugated validated for use in IF, ID & WB.
Research Category
Signaling
Research Sub Category
Growth Factors & Receptors
WORKING DILUTION

The final working dilution is dependent on the specific assay conditions. However, in fluorescence microscopy good results can be expected with dilutions in the following ranges:

1. Incident light excitation

Direct immunofluorescence: 1/10 - 1/80

Indirect immunofluorescence: 1/20 - 1/400

2. Transmitted light excitation

Direct immunofluorescence: 1/5 - 1/40

Indirect immunofluorescence: 1/10 - 1/80

Physical form

The antibody is supplied in 20mM Tris, 70mM NaCl, pH 8.0, with 0.1% sodium azide as preservative.

Storage and Stability

Store at +2 to +8°C, for up to 6 months, undiluted. Protect from light.

WARNING: This reagent contains 0.1% sodium azide as preservative. Due to potential hazards arising from the build up of this material in pipes, spent reagent should be disposed of with liberal volumes of water.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service