CLS3022P2L
Corning® reusable graduated cylinder
single metric scale with funnel top, polypropylene, "to contain", size 2 L
Synonym(s):
lab equipment accessory
Select a Size
€224.00
Select a Size
About This Item
€224.00
Recommended Products
material
polypropylene
feature
graduations 20 mL
manufacturer/tradename
Corning 3022P-2L
H × O.D.
482 mm × 84 mm
size
2 L
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service